Orders LinkContactUsLinkComponent
VWR®, Work Jackets
공급사:   VWR Collection
VWR®, Work Jackets
공급사:  VWR Collection
Certificates
VWR®, Work Jackets
21 Options Available Below
Product Details & Documents
Durable jackets made of 65% polyester and 35% sanforized cotton, which prevents the clothing from shrinking in the laundry.
  • Two breast pockets with press stud, mobile phone pocket, two side pockets, three inside pockets
  • Press stud closure
  • Sleeves and jacket bottom are adjustable
  • Back folds for improved freedom of movement
Weight: 245 g/m²
Specifications for Product Family
사양
  • Catalog No:
  • 113-06729
  • 113-06730
  • 113-06731
  • 113-06732
  • 113-06733
  • 113-06734
  • 113-06735
  • 113-07365
  • 113-07366
  • 113-07367
  • 113-07368
  • 113-07369
  • 113-07370
  • 113-07371
  • 113-07372
  • 113-07373
  • 113-07374
  • 113-07375
  • 113-07376
  • 113-07377
  • 113-07378
  • Garment application:
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Industrial
  • Gender:
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • For men
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Closure type:
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Fabric:
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • 65% polyester and 35% sanforised cotton
  • Weight:
  • 245 g/m²
  • 245 g/m²
  • 245 g/m²
  • 245 g/m²
  • 245 g/m²
  • 245 g/m²
  • 245 g/m²
  • 245 g/m²
  • 245 g/m²
  • 245 g/m²
  • 245 g/m²
  • 245 g/m²
  • 245 g/m²
  • 245 g/m²
  • 245 g/m²
  • 245 g/m²
  • 245 g/m²
  • 245 g/m²
  • 245 g/m²
  • 245 g/m²
  • 245 g/m²
사양
Product Family Options
제품 정보
  • Colour
    의류 사이즈
    재고 여부
    Price
  • Blue/grey
    46 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-06729
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Blue/grey
    48 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-06730
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Blue/grey
    50 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-06731
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Blue/grey
    52 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-06732
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Blue/grey
    54 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-06733
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Blue/grey
    56 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-06734
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Blue/grey
    58 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-06735
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Grey/black
    46 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-07365
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Grey/black
    48 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-07366
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Grey/black
    50 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-07367
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Grey/black
    52 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-07368
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Grey/black
    54 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-07369
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Grey/black
    56 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-07370
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Grey/black
    58 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-07371
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Blue/dark blue
    46 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-07372
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Blue/dark blue
    48 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-07373
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Blue/dark blue
    50 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-07374
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Blue/dark blue
    52 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-07375
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Blue/dark blue
    54 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-07376
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Blue/dark blue
    56 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-07377
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²
  • Blue/dark blue
    58 (men)
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
    사양:
    More
    Specifications
    Cat. No.
    113-07378
    Garment application
    Industrial
    Gender
    For men
    Sleeve or leg length
    Long
    Closure type
    Press studs / snaps
    Fabric
    65% polyester and 35% sanforised cotton
    Weight
    245 g/m²