Orders LinkContactUsLinkComponent
VWR® Redox Electrodes
VWR® Redox Electrodes
공급사:   VWR Collection
VWR® Redox Electrodes
공급사:  VWR Collection
Certificates
VWR® Redox Electrodes
VWR® Redox Electrodes
2 Options Available Below
Product Family Options
제품 정보
  • Connection
    Electrolyte
    Material
    직경×길이
    재고 여부
    Price
  • 662-1408
    RD 113 (redox)
    RD 113 (redox)
    BNC, 1 m cable
    Gel
    Epoxy
    12×120 mm
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
  • 662-1409
    RD 223 (redox)
    RD 223 (redox)
    BNC, 1 m cable
    Gel
    Glass
    12×120 mm