Orders LinkContactUsLinkComponent
장갑 라이너
장갑 라이너
장갑 라이너
장갑 라이너
내구성 있고 재사용 가능한 장갑 라이너는 100% 나일론으로 만들어졌습니다. 엄격한 클린룸 요구 사항과 호환되며 가운 장갑으로 효과적으로 사용할 수 있습니다. 보풀이 적습니다.
  • 장시간 사용시 땀을 감소시키고 수분을 흡수
  • 촉각 감소 없이 피부 자극 방지
  • 장갑의 수명을 연장하고 자원 절약을 위해 마모 및 찢어짐 제한
  • 사전 세탁, 세탁시 수축되지 않음
기본 중량: 21,5 g/pair ±10%
두께: 2 mm ±10%
표면 저항: 1×10⁹ ±10% ohms/m²
Used glove liners can be recycled after use using the Recycling solution for disposable gloves, Zero Waste Box™from TerraCycle.
  • Catalog No:
  • 112-1900
  • 112-1915
  • 112-1899
  • 112-1910
  • 112-1898
  • 112-1905
  • 112-1904
  • 112-1917
  • 112-4326
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
Product Family Options
  • 모델
    장갑 사이즈
    Colour
    두께
    Packed
  • Full-finger liners
    S
    Green
    2mm ± 10%
    20 pairs/bag
    1 * 20 PAIR
    1 * 20 PAIR
    Specifications
    Cat. No.
    112-1900
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Reusable
    Cuff style
    Knit wrist
  • Half-finger liners
    S
    Green
    2mm ± 10%
    20 pairs/bag
    1 * 20 PAIR
    1 * 20 PAIR
    Specifications
    Cat. No.
    112-1915
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Reusable
    Cuff style
    Knit wrist
  • Full-finger liners
    M
    Orange
    2mm ± 10%
    20 pairs/bag
    1 * 20 PAIR
    1 * 20 PAIR
    Specifications
    Cat. No.
    112-1899
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Reusable
    Cuff style
    Knit wrist
  • Half-finger liners
    M
    Orange
    2mm ± 10%
    20 pairs/bag
    1 * 20 PAIR
    1 * 20 PAIR
    Specifications
    Cat. No.
    112-1910
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Reusable
    Cuff style
    Knit wrist
  • Full-finger liners
    L
    Blue
    2mm ± 10%
    20 pairs/bag
    1 * 20 PAIR
    1 * 20 PAIR
    Specifications
    Cat. No.
    112-1898
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Reusable
    Cuff style
    Knit wrist
  • Half-finger liners
    L
    Blue
    2mm ± 10%
    20 pairs/bag
    1 * 20 PAIR
    1 * 20 PAIR
    Specifications
    Cat. No.
    112-1905
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Reusable
    Cuff style
    Knit wrist
  • Full-finger liners
    XL
    Red
    2mm ± 10%
    20 pairs/bag
    1 * 20 PAIR
    1 * 20 PAIR
    Specifications
    Cat. No.
    112-1904
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Reusable
    Cuff style
    Knit wrist
  • Half-finger liners
    XL
    Red
    2mm ± 10%
    20 pairs/bag
    1 * 20 PAIR
    1 * 20 PAIR
    Specifications
    Cat. No.
    112-1917
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Reusable
    Cuff style
    Knit wrist
  • Full-finger liners
    XXL
    Purple
    2mm ± 10%
    20 pairs/bag
    1 * 20 PAIR
    1 * 20 PAIR
    Specifications
    Cat. No.
    112-4326
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Reusable
    Cuff style
    Knit wrist