Robotic tips for Tecan®
49003-1106 (red tray, blister-2), 1000 µl, +, +, Black, Tecan and other OEM workstations, 12 × 2 racks of 96 tips
제품 번호 RITT49003-1106
공급사: RITTER
Jump to:
Product Details & Documents
Black conductive PP or clear PP, with or without filter, sterile or non sterile.
- Free of RNase, DNase, DNA and pyrogens
- Ideal for dosing even small volumes with high precision and without contamination
- Conductive tips: Conductivity enables the system to recognise the filling height and will guarantee a minimal immersion of the tip into the liquid, in order to assure a save pipetting and dispensing
- Clear tips: Offer optical surveillance for the pipetting process (e.g. recognition of foam)
Type 'Tecan' is compatible with the following robotic instruments:
Tecan: Genesis, Freedom EVO, MiniPrep, Cavro
Qiagen/Corbett: CAS 1200/4200/4800, QIAgility
Perkin Elmer (Packard): Multiprobe II, Janus (with Multiprobe head)
Abbott Diagnostic: m2000
Documents
Ritter Pure clean origin - Robotic consumables
Product Literature
사양
- Packed:12 × 2 racks of 96 tips
- Sterile:+
- 용량:1000 µl
- Seq:11
- Tip style:49003-1106 (red tray, blister-2)
- Tip colour:Black
- 사용 대상:Tecan and other OEM workstations
- Filter:+