VWR® IPASEPT® 70
IPASEPT® 70, 6×1 L trigger spray bottles, Plastic bottle
공급사: VWR Chemicals
Jump to:
Product Details & Documents
Iso-propyl alcohol Ph. Eur. 70% v/v, with purified water Ph. Eur. 30% v/v, filtered at 0,2 μm.
Filled under GMP conditions in cleanroom ISO Class 7
-
Certificate of Analysis available for each batch
For surface spray cleaning, with or without wipes. Also for cleaning gloved hands before entry into cleanrooms. Ideal for wiping down entrances to controlled environments.
Biocide. Disinfectant.
주의: Caution: Use biocides safely. Always read the label and product information before use.
사양
- 설명:IPASEPT® 70
- 포장 유형:Plastic bottle
- Packed:6×1 L trigger spray bottles