Orders LinkContactUsLinkComponent
VWR®, Bouffant Caps, PP, Detectable
VWR®, Bouffant Caps, PP, Detectable
Supplier:   VWR Collection
VWR®, Bouffant Caps, PP, Detectable
Supplier:  VWR Collection
Certificates
VWR®, Bouffant Caps, PP, Detectable
2 Options Available Below
Product Details & Documents
These detectable caps, made of non woven polypropylene, provide an effective way to cover the hair when working in hygienic environments.
  • Increased detectability thanks to foil strip
  • Two elastic bands for improved fit
Weight: 10 g/m².
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-1578
  • 113-1579
  • Garment application:
  • Laboratory
  • Laboratory
Specifications
Product Family Options
Product Information
  • Colour
    직경
    Availability
    Price
  • Blue
    530 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-1578
    Garment application
    Laboratory
  • Blue
    610 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-1579
    Garment application
    Laboratory