Orders LinkContactUsLinkComponent
2-Propanol ≥98%, TECHNICAL
2-Propanol ≥98%, TECHNICAL
Flammable
Irritant
   Sustainable Options Available
Supplier:   VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
2-Propanol ≥98%, TECHNICAL
6 Options Available Below
Product Details & Documents
Synonyms:  IPA, Isopropyl Rubbing Alcohol, Isopropyl alcohol, Dimethyl carbinol, 2-Hydroxy propane, Isopropanol, Propan-2-ol
  • CAS Number:67-63-0
  • Formula:(CH₃)₂CHOH
  • MDL Number:MFCD00011674
  • Molecular Weight:60.1 g/mol
  • Merck Index:13,05228
  • Boiling Pt:82 °C (1013 hPa)
  • Melting Pt:–89 °C
  • Density:0,786 g/cm³ (20 °C)
  • Flash Pt:12 °C
  • Storage Temperature:Ambient
  • EINECS:200-661-7
  • UN:1219
  • ADR:3,II
  • REACH:01-2119457558-25
Specifications for Product Family
Specification Test Results
  • Assay:
    Min. 98 %
Specifications
Specifications
  • Catalog No:
  • 20922.320
  • 20922.364
  • 20922.368
  • 20922.411
  • 20922.466
  • 20922.557
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Product Family Options
Product Information
  • 포장 유형
    Pk
    Availability
    Price
  • Plastic bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - ₩73,357
    ₩73,357
    1 * 2,5 L
       ₩73,357
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    20922.320
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
      Safer Human and Environmental Health
  • Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - ₩125,150
    ₩125,150
    1 * 5 L
       ₩125,150
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    20922.364
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
      Safer Human and Environmental Health
  • Canister
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - ₩172,671
    ₩172,671
    1 * 5 L
       ₩172,671
    Specifications:
    More
    Specifications
    Cat. No.
    20922.368
    Environmentally Preferable
    Environmental certification
  • Plastic drum
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    20922.411
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
      Safer Human and Environmental Health
  • Plastic drum
    25 L
    Log in for availability
    Log in for availability
    1 * 25 L
    1 * 25 L
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    20922.466
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
      Safer Human and Environmental Health
  • Metal drum
    200 L
    Log in for availability
    Log in for availability
    1 * 200 L
    1 * 200 L
    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    20922.557
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
      Safer Human and Environmental Health