Sodium hydroxide concentrate for dilution to 0.1 N in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:∼105 °C (1013 hPa)
Density:1,327 g/cm³ (20 °C)
EINECS:215-185-5
UN:1824
ADR:8,II
:Management of Change (MOC) category = N
REACH:01-2119457892-27
:GHS05
Storage Temperature:Ambient
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:0.0998-0.1002
- Normality (by titrimetry):Passes Test