클린룸 작업복, VWR® Maximum, SF
VWR® Maximum SF overalls are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and coated with breathable polyethylene film, these garments have been designed to optimise barrier protection, cleanliness and comfort properties.
- Available as a standard overall with collar, either elasticated ankles or non slip boot covers, or hooded overall with fluid-impervious boot covers
- Inset sleeves, zip closure, elasticated cuffs
- Superior fluid and particulate barrier
- Lowest level of particle shedding: Level 1 Helmke drum classification
- Light chemical splash resistance
- Improved breathability to ensure user comfort
- Soft, cloth-like fabric with maximum tear strength
Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
VWR® Maximum SF overalls are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and coated with breathable polyethylene film, these garments have been designed to optimise barrier protection, cleanliness and comfort properties.
- Available as a standard overall with collar, either elasticated ankles or non slip boot covers, or hooded overall with fluid-impervious boot covers
- Inset sleeves, zip closure, elasticated cuffs
- Superior fluid and particulate barrier
- Lowest level of particle shedding: Level 1 Helmke drum classification
- Light chemical splash resistance
- Improved breathability to ensure user comfort
- Soft, cloth-like fabric with maximum tear strength
Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing.
- Catalog No:
- 113-1078
- 113-1079
- 113-1080
- VWRU414004-311
- 113-1081
- VWRU414004-310
- 113-1082
- 113-1084
- 113-1085
- 113-1086
- 113-1087
- 113-1088
- 113-1089
- VWRU414004-686
- 113-9161
- 113-1074
- 113-1075
- VWRU414004-308
- 113-1076
- 113-1077
- VWRU414004-683
- VWRU414004-684
- VWRU414004-685
- Model:
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- SF
- Garment application:
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Bacterial filtration efficiency:
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
- 99,9%
Product Family Options
Standard overall with collar
- Colour의류 사이즈
Specifications
Cat. No.113-1078ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.113-1079ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.113-1080ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.VWRU414004-311ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.113-1081ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.VWRU414004-310ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.113-1082ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.113-1084ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%
Overall with collar and attached non skid boot covers
- Colour의류 사이즈
Specifications
Cat. No.113-1085ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.113-1086ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.113-1087ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.113-1088ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.113-1089ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.VWRU414004-686ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%
Hooded overall with attached fluid-impervious boot covers
- Colour의류 사이즈
Specifications
Cat. No.113-9161ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.113-1074ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.113-1075ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.VWRU414004-308ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.113-1076ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.113-1077ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.VWRU414004-683ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.VWRU414004-684ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%Specifications
Cat. No.VWRU414004-685ModelSFGarment applicationCleanroom|LaboratoryBacterial filtration efficiency99,9%