VWR® Basic Frocks, SMS
공급사: VWR Collection
이 제품에 관하여
These SMS frocks are manufactured from a latex-free, multi-layer spunbonded - melt-blown - spunbonded (SMS) fabric, that offers high tensile strength while remaining soft, comfortable and breathable.
- Snap collar, raglan sleeves, snap closure
- Fluid and particulate barrier
- Light chemical splash resistance
Colour: White or blue
11 Options Available Below
Product Details & Documents
These SMS frocks are manufactured from a latex-free, multi-layer spunbonded - melt-blown - spunbonded (SMS) fabric, that offers high tensile strength while remaining soft, comfortable and breathable.
- Snap collar, raglan sleeves, snap closure
- Fluid and particulate barrier
- Light chemical splash resistance
Colour: White or blue
Specifications for Product Family
사양
- Catalog No:
- 113-1156
- 113-1157
- 113-1158
- 113-1159
- 113-1160
- 113-1161
- 113-1162
- 113-1174
- 113-1175
- 113-1176
- 113-1166
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Pockets:
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Garment application:
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
사양
Product Family Options
제품 정보
- Colour의류 사이즈재고 여부Price
- 사양:
More
Specifications
Cat. No.113-1156Closure typePress studs / snapsPocketsWithout pocketsGenderUnisexGarment applicationCleanroom|Laboratory - 사양:
More
Specifications
Cat. No.113-1157Closure typePress studs / snapsPocketsWithout pocketsGenderUnisexGarment applicationCleanroom|Laboratory - 사양:
More
Specifications
Cat. No.113-1158Closure typePress studs / snapsPocketsWithout pocketsGenderUnisexGarment applicationCleanroom|Laboratory - 사양:
More
Specifications
Cat. No.113-1159Closure typePress studs / snapsPocketsWithout pocketsGenderUnisexGarment applicationCleanroom|Laboratory - 사양:
More
Specifications
Cat. No.113-1160Closure typePress studs / snapsPocketsWithout pocketsGenderUnisexGarment applicationCleanroom|Laboratory - 사양:
More
Specifications
Cat. No.113-1161Closure typePress studs / snapsPocketsWithout pocketsGenderUnisexGarment applicationCleanroom|Laboratory - 사양:
More
Specifications
Cat. No.113-1162Closure typePress studs / snapsPocketsWithout pocketsGenderUnisexGarment applicationCleanroom|Laboratory - 사양:
More
Specifications
Cat. No.113-1174Closure typePress studs / snapsPocketsWithout pocketsGenderUnisexGarment applicationCleanroom|Laboratory - 사양:
More
Specifications
Cat. No.113-1175Closure typePress studs / snapsPocketsWithout pocketsGenderUnisexGarment applicationCleanroom|Laboratory - 사양:
More
Specifications
Cat. No.113-1176Closure typePress studs / snapsPocketsWithout pocketsGenderUnisexGarment applicationCleanroom|Laboratory - 사양:
More
Specifications
Cat. No.113-1166Closure typePress studs / snapsPocketsWithout pocketsGenderUnisexGarment applicationCleanroom|Laboratory