Orders LinkContactUsLinkComponent
VWR® Basic Beard Covers, SPP
VWR® Basic Beard Covers, SPP
공급사:  VWR Collection
Certificates
VWR® Basic Beard Covers, SPP
VWR® Basic Beard Covers, SPP
3 Options Available Below
Product Details & Documents
VWR® Basic SPP beard covers are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Lightweight and breathable first line barrier protection
This garment is ideal for less critical areas or pre-gowning entry rooms. It is tested and manufactured in an ISO certified facility, lot-controlled and validated through independent laboratory testing.
Diameter (stretched): 406 mm
Product Family Options
제품 정보
  • Colour
    재고 여부
    Price
  • 113-1463
    With latex
    With latex
    White
    Log in 재고 확인
    Log in 재고 확인
    1 * 1.000 items
    1 * 1.000 items
  • Latex-free
    White
    Log in 재고 확인
    Log in 재고 확인
    1 * 1.000 items
    1 * 1.000 items
  • VWRU89107-778
    Latex-free
    Latex-free
    White
    Log in 재고 확인
    Log in 재고 확인
    1 * 1.000 items
    1 * 1.000 items