VWR® Basic Cleanroom Overalls, SPP
이 제품에 관하여
VWR® Basic SPP overalls are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Tapered collar, inset sleeves, zip closure, elasticated cuffs
- Lightweight and breathable first line barrier protection
This garment is ideal for less critical areas or pre-gowning entry rooms.
Tested and manufactured in an ISO certified facility also lot-controlled and validated through independent laboratory testing.
7 Options Available Below
Product Details & Documents
VWR® Basic SPP overalls are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
- Tapered collar, inset sleeves, zip closure, elasticated cuffs
- Lightweight and breathable first line barrier protection
This garment is ideal for less critical areas or pre-gowning entry rooms.
Tested and manufactured in an ISO certified facility also lot-controlled and validated through independent laboratory testing.
Specifications for Product Family
사양
- Catalog No:
- 113-1104
- 113-1105
- 113-1106
- 113-1107
- 113-1108
- 113-1109
- 113-1110
- Garment application:
- Cleanroom|General Purpose|Laboratory
- Cleanroom|General Purpose|Laboratory
- Cleanroom|General Purpose|Laboratory
- Cleanroom|General Purpose|Laboratory
- Cleanroom|General Purpose|Laboratory
- Cleanroom|General Purpose|Laboratory
- Cleanroom|General Purpose|Laboratory
사양
Product Family Options
제품 정보
- Colour의류 사이즈재고 여부Price
- 113-1104Without hood사양:
More
Specifications
Cat. No.113-1104Garment applicationCleanroom|General Purpose|Laboratory - 113-1105Without hood사양:
More
Specifications
Cat. No.113-1105Garment applicationCleanroom|General Purpose|Laboratory - 113-1106Without hood사양:
More
Specifications
Cat. No.113-1106Garment applicationCleanroom|General Purpose|Laboratory - 113-1107Without hood사양:
More
Specifications
Cat. No.113-1107Garment applicationCleanroom|General Purpose|Laboratory - 113-1108Without hood사양:
More
Specifications
Cat. No.113-1108Garment applicationCleanroom|General Purpose|Laboratory - 113-1109Without hood사양:
More
Specifications
Cat. No.113-1109Garment applicationCleanroom|General Purpose|Laboratory - 113-1110Without hood사양:
More
Specifications
Cat. No.113-1110Garment applicationCleanroom|General Purpose|Laboratory