Orders LinkContactUsLinkComponent
VWR® Basic Cleanroom Lab Coats, SPP, Disposable
VWR® Basic Cleanroom Lab Coats, SPP, Disposable
공급사:  VWR Collection
Certificates
VWR® Basic Cleanroom Lab Coats, SPP, Disposable
26 Options Available Below
Product Details & Documents
VWR® Basic SPP lab coats are manufactured from a strong, lightweight and breathable spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates in non hazardous environments.
  • Tapered collar, inset sleeves, snap closure
  • Elasticated or knitted cuffs, no pockets or three pockets
  • Lightweight and breathable first line barrier protection
This garment is ideal for less critical areas or pre-gowning entry rooms. It is tested and manufactured in an ISO certified facility and lot controlled and validated through independent laboratory testing.
 : 25 or 30 coats per carton.
Specifications for Product Family
사양
  • Catalog No:
  • 113-1118
  • 113-1119
  • VWRU414004-353
  • 113-1120
  • VWRU414004-351
  • 113-1121
  • VWRU414004-350
  • 113-1122
  • VWRU89136-824
  • 113-1123
  • 113-1124
  • VWRU414004-354
  • VWRU89136-828
  • 113-1111
  • VWRU414004-346
  • 113-1112
  • VWRU414004-345
  • 113-1113
  • VWRU414004-344
  • VWRU89208-750
  • 113-1114
  • VWRU414004-342
  • 113-1115
  • VWRU414004-343
  • 113-1116
  • VWRU414004-348
  • Closure type:
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Pockets:
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • Without pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Garment application:
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Cleanroom|Laboratory|Industrial
  • Weight:
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
  • 45 g/m²
사양
Product Family Options
With elasticated cuffs, no pockets
제품 정보
  • Colour
    의류 사이즈
    길이
    재고 여부
    Price
  • White
    S
    102 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 25 items
    1 * 25 items
    사양:
    More
    Specifications
    Cat. No.
    113-1118
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    M
    102 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 25 items
    1 * 25 items
    사양:
    More
    Specifications
    Cat. No.
    113-1119
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    M
    102 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 25 items
    1 * 25 items
    사양:
    More
    Specifications
    Cat. No.
    VWRU414004-353
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    L
    102 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 25 items
    1 * 25 items
    사양:
    More
    Specifications
    Cat. No.
    113-1120
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    L
    102 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 25 items
    1 * 25 items
    사양:
    More
    Specifications
    Cat. No.
    VWRU414004-351
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    XL
    110 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 25 items
    1 * 25 items
    사양:
    More
    Specifications
    Cat. No.
    113-1121
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    XL
    102 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 25 items
    1 * 25 items
    사양:
    More
    Specifications
    Cat. No.
    VWRU414004-350
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    XXL
    110 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 25 items
    1 * 25 items
    사양:
    More
    Specifications
    Cat. No.
    113-1122
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    XXL
    Log in 재고 확인
    Log in 재고 확인
    1 * 30 items
    1 * 30 items
    사양:
    More
    Specifications
    Cat. No.
    VWRU89136-824
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    3XL
    110 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 25 items
    1 * 25 items
    사양:
    More
    Specifications
    Cat. No.
    113-1123
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    4XL
    110 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 25 items
    1 * 25 items
    사양:
    More
    Specifications
    Cat. No.
    113-1124
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    4XL
    Log in 재고 확인
    Log in 재고 확인
    1 * 25 items
    1 * 25 items
    사양:
    More
    Specifications
    Cat. No.
    VWRU414004-354
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    5XL
    Log in 재고 확인
    Log in 재고 확인
    1 * 30 items
    1 * 30 items
    사양:
    More
    Specifications
    Cat. No.
    VWRU89136-828
    Closure type
    Press studs / snaps
    Pockets
    Without pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
With knitted cuffs, three pockets
제품 정보
  • Colour
    의류 사이즈
    길이
    재고 여부
    Price
  • White
    S
    100 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 30 items
    1 * 30 items
    사양:
    More
    Specifications
    Cat. No.
    113-1111
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    S
    100 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 30 items
    1 * 30 items
    사양:
    More
    Specifications
    Cat. No.
    VWRU414004-346
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    M
    100 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 30 items
    1 * 30 items
    사양:
    More
    Specifications
    Cat. No.
    113-1112
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    M
    100 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 30 items
    1 * 30 items
    사양:
    More
    Specifications
    Cat. No.
    VWRU414004-345
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    L
    125 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 30 items
    1 * 30 items
    사양:
    More
    Specifications
    Cat. No.
    113-1113
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    L
    100 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 30 items
    1 * 30 items
    사양:
    More
    Specifications
    Cat. No.
    VWRU414004-344
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • Blue
    L
    Log in 재고 확인
    Log in 재고 확인
    1 * 30 items
    1 * 30 items
    사양:
    More
    Specifications
    Cat. No.
    VWRU89208-750
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    XL
    102 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 30 items
    1 * 30 items
    사양:
    More
    Specifications
    Cat. No.
    113-1114
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    XL
    Log in 재고 확인
    Log in 재고 확인
    1 * 30 items
    1 * 30 items
    사양:
    More
    Specifications
    Cat. No.
    VWRU414004-342
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    XXL
    100 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 30 items
    1 * 30 items
    사양:
    More
    Specifications
    Cat. No.
    113-1115
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    XXL
    100 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 30 items
    1 * 30 items
    사양:
    More
    Specifications
    Cat. No.
    VWRU414004-343
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    3XL
    100 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 30 items
    1 * 30 items
    사양:
    More
    Specifications
    Cat. No.
    113-1116
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²
  • White
    3XL
    100 cm
    Log in 재고 확인
    Log in 재고 확인
    1 * 30 items
    1 * 30 items
    사양:
    More
    Specifications
    Cat. No.
    VWRU414004-348
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    Cleanroom|Laboratory|Industrial
    Weight
    45 g/m²