Orders LinkContactUsLinkComponent
VWR®, Electrode cleaning solutions
VWR®, Electrode cleaning solutions
VWR®, Electrode cleaning solutions
공급사:  VWR Chemicals
Certificates
VWR®, Electrode cleaning solutions
VWR®, Electrode cleaning solutions
3 Options Available Below
Product Family Options
제품 정보
  • 포장 유형
    Pk
    재고 여부
    Price
  • 83603.180
    Electrode cleaning solution pepsin/hydrochloric acid for removal of proteins
    Electrode cleaning solution pepsin/hydrochloric acid for removal of proteins
    Plastic bottle
    100 mL
    Log in 재고 확인
    Log in 재고 확인
    1 * 100 mL
    1 * 100 mL - ₩50,328
    ₩50,328
    1 * 100 mL
       ₩50,328
  • 83603.260
    Electrode cleaning solution pepsin/hydrochloric acid for removal of proteins
    Electrode cleaning solution pepsin/hydrochloric acid for removal of proteins
    Plastic bottle
    500 mL
    Log in 재고 확인
    Log in 재고 확인
    1 * 500 mL
    1 * 500 mL - ₩140,939
    ₩140,939
    1 * 500 mL
       ₩140,939
  • 83603.290
    Electrode cleaning solution pepsin/hydrochloric acid for removal of proteins
    Electrode cleaning solution pepsin/hydrochloric acid for removal of proteins
    Plastic bottle
    1 L
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 L
    1 * 1 L - ₩224,722
    ₩224,722
    1 * 1 L
       ₩224,722