VWR®, Draining Racks
Drying rack, 457×181×525 mm
제품 번호 211-0196
공급사: VWR Collection
Jump to:
Product Details & Documents
Wire, vinyl coated.
- Can be used on the bench or as a wall rack
- Drip tray can be removed from the side for easy emptying
Has 18 draining rods, 127 mm long for large containers and 72 draining rods, 76 mm long for smaller containers.
사양
- 설명:Drying rack
- W×D×H:457×181×525 mm