VWR®, 진공/압력 벌브(bulb)
튜브가 없는 진공/압력 벌브
제품 번호 612-2705
공급사: VWR Collection
Jump to:
Product Details & Documents
Black rubber with PVC connectors.
- For collecting gas-phase samples or aspirating gases through instruments
- Two thick-walled rubber valves
용량: 85 ml
Colour: black
주문 정보: Supplied without tubing.
Documents
사양
- 설명:튜브가 없는 진공/압력 벌브