Orders LinkContactUsLinkComponent
VWR®, Laboratory Jacks
공급사:   VWR Collection
VWR®, Laboratory Jacks
공급사:  VWR Collection
Certificates
VWR®, Laboratory Jacks
VWR®, Laboratory Jacks
6 Options Available Below
Product Details & Documents
Anodised aluminium. With M10 threaded hole in the middle of the top plate for attaching retort rods (starting from item number 245-0006).
  • Smooth adjustment of working height
  • Extreme stability
  • Exceptionally sturdy
  • Corrosion resistant and non susceptible to a wide range of chemicals
Product Family Options
제품 정보
  • 플레이트 크기
    Max. static load capacity
    Working height
    Colour
    재고 여부
    Price
  • 50×40 mm
    20 kg
    31 - 93 mm
    Green
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
  • 80×75 mm
    30 kg
    49 - 147 mm
    Blue
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
  • 120×140 mm
    50 kg
    60 - 243 mm
    Pink
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
  • 122×150 mm
    80 kg
    80 - 278 mm
    Yellow
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
  • 180×216 mm
    90 kg
    95 - 398 mm
    Blue
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
  • 240×320 mm
    90 kg
    120 - 500 mm
    Grey