Orders LinkContactUsLinkComponent
VWR®, 사각 냉동 라벨
VWR®, 사각 냉동 라벨
Supplier:   VWR Collection
VWR®, 사각 냉동 라벨
Supplier:  VWR Collection
Certificates
VWR®, 사각 냉동 라벨
VWR®, 사각 냉동 라벨
10 Options Available Below
Product Details & Documents
라벨과 접착제는 –196 ~ +121 ° C의 온도를 견딜 수 있도록 설계되었습니다. VWR® 라벨은 모든 냉동 보관에 이상적이며 거의 모든 종류의 인큐베이터에서 사용할 수 있습니다.
  • 코팅할 필요가 없습니다
  • 튜브, 랙 및 박스에 사용
Documents
Product Family Options
Product Information
  • 설명
    Printing method
    Colour
    치수
    Packed
    Availability
    Price
  • 레이저 프린터에 적합한 1,5 ml 튜브용 라벨
    Laser
    White
    33×13 mm
    Log in for availability
    Log in for availability
    1 * 1.700 items
    1 * 1.700 items
  • Labels for 1,5 - 2 ml tubes, on roll
    White
    33×13 mm
    On roll
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
  • Printer labels for racks and boxes, suitable for Printer labelss
    Laser
    White
    67×25 mm
    Log in for availability
    Log in for availability
    1 * 600 items
    1 * 600 items
  • Printer labels for 1,5 ml tubes
    Laser
    Multicolour
    24×13 mm
    Log in for availability
    Log in for availability
    1 * 2.125 items
    1 * 2.125 items
  • Printer labels for 0,5 ml tubes
    Laser
    White
    24×13 mm
    Log in for availability
    Log in for availability
    1 * 2.380 items
    1 * 2.380 items
  • 0.2ml PCR 튜브용 라벨, 롤 용지
    White
    20×6 mm
    On roll
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
  • Labels for 1,5 - 2 ml tubes, on roll
    Yellow
    33×13 mm
    On roll
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
  • Labels for 1,5 - 2 ml tubes, on roll
    Blue
    33×13 mm
    On roll
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
  • Labels for 1,5 - 2 ml tubes, on roll
    Red
    33×13 mm
    On roll
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items
  • Labels for 1,5 - 2 ml tubes, on roll
    Green
    33×13 mm
    On roll
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items