Orders LinkContactUsLinkComponent
VWR®, 슈커버, LDPE/LLDPE
VWR®, 슈커버, LDPE/LLDPE
VWR®, 슈커버, LDPE/LLDPE
Supplier:  VWR Collection
Certificates
VWR®, 슈커버, LDPE/LLDPE
2 Options Available Below
Product Details & Documents
65% 저밀도 PE(LDPE)와 35% 선형 저밀도 PE(LLDPE)를 혼합한 CPE로 만든 슈커버입니다.
  • 습기와 먼지에 대한 방수제
  • 발 아래 이음새 없음
L×H: 410×150 mm
* One size fits all
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-8281
  • 113-8284
  • Garment application:
  • Laboratory
  • Laboratory
Specifications
Product Family Options
Product Information
  • Colour
    신발 사이즈
    Availability
    Price
  • Blue
    U*
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8281
    Garment application
    Laboratory
  • White
    U*
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8284
    Garment application
    Laboratory