Orders LinkContactUsLinkComponent
VWR®, 바이저(visor) 포함 라운드 캡(Bouffant cap), PP
VWR®, 바이저(visor) 포함 라운드 캡(Bouffant cap), PP
Supplier:   VWR Collection
VWR®, 바이저(visor) 포함 라운드 캡(Bouffant cap), PP
Supplier:  VWR Collection
Certificates
VWR®, 바이저(visor) 포함 라운드 캡(Bouffant cap), PP
VWR®, 바이저(visor) 포함 라운드 캡(Bouffant cap), PP
3 Options Available Below
Product Details & Documents
Bouffant caps made from non woven PP. For non critical applications.
  • With visor (white or blue)
  • Latex-free
  • Available with mesh cap for use with long hair
Weight: 12 g/m².
Packaging: 백으로 제공.
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-8251
  • 113-8252
  • 113-8253
  • Garment application:
  • Laboratory
  • Laboratory
  • Laboratory
Specifications
Product Family Options
Product Information
  • 설명
    Colour
    직경
    Availability
    Price
  • Standard
    White
    510 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8251
    Garment application
    Laboratory
  • Standard
    Blue
    510 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8252
    Garment application
    Laboratory
  • For long hair
    White
    510 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8253
    Garment application
    Laboratory