VWR®, Rack for Slide Boxes
Stackable drawer, Acrylic, 50/100, 227×264×118 mm
제품 번호 631-0742
공급사: VWR Collection
Jump to:
Product Details & Documents
Storage racks for microscopy slides make organisation of samples easy.
- Clear acrylic allows the labels on the slide boxes to be seen
- Boxes can be placed in rows or columns
- Sorted by colour to create an easy coding system
Non skid rubber feet allow for secure bench top placement, whilst pre-drilled holes enable under the counter or wall mounting.
사양
- No. of slides:50/100
- 설명:Stackable drawer
- Material:Acrylic
- W×D×H:227×264×118 mm