Orders LinkContactUsLinkComponent
VWR® Wire-O Computation Notebooks
VWR® Wire-O Computation Notebooks
공급사:   VWR Collection
VWR® Wire-O Computation Notebooks
공급사:  VWR Collection
Certificates
VWR® Wire-O Computation Notebooks
VWR® Wire-O Computation Notebooks
5 Options Available Below
Product Details & Documents
Wire-O computation notebooks feature a translux cover and quad ruled format pages.
  • Translux cover
  • 235×298,5 mm pages
  • 100 pages
  • Quad ruled format
Certifications: All books made in accordance with GLP, GMP, and GCP guidelines.
Product Family Options
제품 정보
  • Colour
    Type
    Version
    Paper size
    No. of pages
    재고 여부
    Price
  • 818-0306
    Wire-O computation notebook
    Wire-O computation notebook
    Translucent
    Quad ruled
    English
    23.5×29.85 cm
    100
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
  • 818-0324
    Wire-O computation notebook
    Wire-O computation notebook
    Translucent
    Quad ruled
    English-German
    23.5×29.85 cm
    100
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
  • 818-0325
    Wire-O computation notebook
    Wire-O computation notebook
    Translucent
    Quad ruled
    English-French
    23.5×29.85 cm
    100
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
  • 818-0326
    Wire-O computation notebook
    Wire-O computation notebook
    Translucent
    Quad ruled
    English-Italian
    23.5×29.85 cm
    100
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
  • 818-0327
    Wire-O computation notebook
    Wire-O computation notebook
    Translucent
    Quad ruled
    English-Spanish
    23.5×29.85 cm