Orders LinkContactUsLinkComponent
VWR® PHP Pipettes, Special Edition
VWR® PHP Pipettes, Special Edition
Supplier:   VWR Collection
VWR® PHP Pipettes, Special Edition
Supplier:  VWR Collection
Certificates
VWR® PHP Pipettes, Special Edition
VWR® PHP Pipettes, Special Edition
35 Options Available Below
Product Details & Documents
VWR® PHP pipettes are fully autoclavable, high performance pipettes with low plunger force.
  • Superior accuracy and precision (tested in ISO 17025 accredited lab)
  • Magnet assisted piston for low plunger force and lesser chance of RSI
  • PVDF tip cone for durable performance
  • Large and clear 4 digit display; color coding for easy identification
  • Three-year warranty
VWR® PHP Pipettes are Precise High Performance, fully autoclavable pipettes, that are available in Single and Multi Channel range. VWR® PHP offers one of the lowest plunger forces and a durable tip cone. The tip ejector is made using special corrosion resistant alloy, protecting it against chemical and physical corrosion. The pipette has an effortless tip ejection with unique shock absorbing mechanism. The tip cone is spring loaded for lesser strain and proper tip fitment for precise pipetting (for Multi Channel only).
Additional Informatiom: PHP Pipette, with a shift in the pipette's hue from blue to pink. A change of pink colored pipette finding it’s way in the hands of the scientists. With the power to make a difference in patients suffering from Breast Cancer, there by leading by its core virtue of setting science in motion to create a better world.
Product Family Options
Single channel variable volume
Product Information
  • 용량
    Availability
    Price
  • 0,1 - 2,5 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 0,5 - 10 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 2 - 20 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 5 - 50 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 10 - 100 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 20 - 200 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 100 - 1000 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 1000 - 5000 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 1000 - 10000 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
Single channel fixed volume
Product Information
  • 용량
    Availability
    Price
  • 2,5 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 5 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 10 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 20 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 25 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 50 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 100 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 200 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 250 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 500 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 1000 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 2000 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 5000 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 10000 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
8-channel variable volume
Product Information
  • 용량
    Availability
    Price
  • 0,5 - 10 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 2 - 20 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 5 - 50 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 10 - 100 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 20 - 200 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 30 - 300 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
12-channel variable volume
Product Information
  • 용량
    Availability
    Price
  • 0,5 - 10 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 2 - 20 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 5 - 50 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 10 - 100 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 20 - 200 µl
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items
  • 30 - 300 µl