Orders LinkContactUsLinkComponent
VWR®, Macro Pipette Tips
VWR®, Macro Pipette Tips
VWR®, Macro Pipette Tips
공급사:  VWR Collection
Certificates
VWR®, Macro Pipette Tips
VWR®, Macro Pipette Tips
14 Options Available Below
Product Details & Documents
Improved accuracy over serological pipettes.
  • Tip length facilitates access into bottles, cell culture flasks, and tubes
Universal fit for compatibility with most major pipettor brands.
Certifications: Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free. Sterile product is certified pyrogen-free.
Product Family Options
5 ml macro tips
제품 정보
  • Tip style
    용량
    Sterile
    Filter
    길이
    Tip colour
    사용 대상
    Packed
    재고 여부
    Price
  • Macro pipette tips
    1000 - 5000 µl
    +
    -
    123,1 mm
    Clear
    Eppendorf, Socorex, Sartorius Biohit
    Rack of 50 tips
    Log in 재고 확인
    Log in 재고 확인
    1 * 50 items
    1 * 50 items
  • Macro pipette tips
    1000 - 5000 µl
    +
    +
    123,1 mm
    Clear
    Eppendorf, Socorex, Sartorius Biohit
    Rack of 50 tips
    Log in 재고 확인
    Log in 재고 확인
    1 * 50 items
    1 * 50 items
  • Macro pipette tips
    1000 - 5000 µl
    -
    +
    123,1 mm
    Clear
    Eppendorf, Socorex, Sartorius Biohit
    Bulk, 100 per bag
    Log in 재고 확인
    Log in 재고 확인
    1 * 100 items
    1 * 100 items
  • Macro pipette tips
    1000 - 5000 µl
    -
    -
    123,1 mm
    Clear
    Eppendorf, Socorex, Sartorius Biohit
    Bulk, 250 per bag
    Log in 재고 확인
    Log in 재고 확인
    1 * 250 items
    1 * 250 items
  • Macro pipette tips
    1000 - 5000 µl
    +
    -
    123,6 mm
    Clear
    VWR, Gilson, Rainin Classic, and other universal-style pipettors
    Rack of 50 tips
    Log in 재고 확인
    Log in 재고 확인
    1 * 50 items
    1 * 50 items
  • Macro pipette tips
    1000 - 5000 µl
    +
    +
    123,6 mm
    Clear
    VWR, Gilson, Rainin Classic, and other universal-style pipettors
    Rack of 50 tips
    Log in 재고 확인
    Log in 재고 확인
    1 * 50 items
    1 * 50 items
  • Macro pipette tips
    1000 - 5000 µl
    -
    +
    123,6 mm
    Clear
    VWR, Gilson, Rainin Classic, and other universal-style pipettors
    Bulk, 100 per bag
    Log in 재고 확인
    Log in 재고 확인
    1 * 100 items
    1 * 100 items
  • Macro pipette tips
    1000 - 5000 µl
    -
    -
    123,6 mm
    Clear
    VWR, Gilson, Rainin Classic, and other universal-style pipettors
    Bulk, 250 per bag
    Log in 재고 확인
    Log in 재고 확인
    1 * 250 items
    1 * 250 items
10 ml macro tips
제품 정보
  • Tip style
    용량
    Sterile
    Filter
    길이
    Tip colour
    사용 대상
    Packed
    재고 여부
    Price
  • Macro pipette tips
    5000 - 10000 µl
    +
    -
    143,8 mm
    Blue
    Socorex, Finn
    Rack of 25 tips
    Log in 재고 확인
    Log in 재고 확인
    1 * 25 items
    1 * 25 items
  • Macro pipette tips
    5000 - 10000 µl
    -
    -
    143,8 mm
    Blue
    Socorex, Finn
    Bulk, 100 per bag
    Log in 재고 확인
    Log in 재고 확인
    1 * 100 items
    1 * 100 items
  • Macro pipette tips
    5000 - 10000 µl
    +
    -
    160,8 mm
    Clear
    VWR, Eppendorf, Gilson, Rainin Classic, Biohit, and other universal-style pipettors
    Rack of 25 tips
    Log in 재고 확인
    Log in 재고 확인
    1 * 25 items
    1 * 25 items
  • Macro pipette tips
    5000 - 10000 µl
    +
    +
    160,8 mm
    Clear
    VWR, Eppendorf, Gilson, Rainin Classic, Biohit, and other universal-style pipettors
    Rack of 25 tips
    Log in 재고 확인
    Log in 재고 확인
    1 * 25 items
    1 * 25 items
  • Macro pipette tips
    5000 - 10000 µl
    -
    +
    160,8 mm
    Clear
    VWR, Eppendorf, Gilson, Rainin Classic, Biohit, and other universal-style pipettors
    Bulk, 50 per bag
    Log in 재고 확인
    Log in 재고 확인
    1 * 50 items
    1 * 50 items
  • Macro pipette tips
    5000 - 10000 µl
    -
    -
    160,8 mm
    Clear
    VWR, Eppendorf, Gilson, Rainin Classic, Biohit, and other universal-style pipettors
    Bulk, 100 per bag