J.T.Baker® BAKERBOND® Q2100, New Generation UHPLC and HPLC Columns
Supplier: Avantor
New Product
Certificates
About this item
BAKERBOND® Q2100 LC phases have been specially designed to solve the challenges of most routine samples. They can also be a good starting point for new method development projects. With options of four widely used chemistries and three particle sizes BAKERBOND® Q2100 can be a useful addition to your analytical toolkit.
- High efficiency UHPLC/HPLC columns
- Based on high purity spherical silica
- C18, C8, phenyl-hexyl and silica phases
- Available with 1,8, 3, 5 µm particle sizes
Specifications
- Catalog No:
- 7354-64
- 7378-64
- 7405-64
- 7411-64
- 7354-24
- 7382-26
- 7373-44
- 7382-44
- 7405-24
- 7358-44
- 7375-44
- 7373-24
- 7378-44
- 7405-44
- 7411-44
- 7354-44
- 7358-24
- Phase name:
- C8
- C8
- C18
- C18
- C8
- Phenyl-Hexyl
- Silica
- Phenyl-Hexyl
- C18
- Phenyl-Hexyl
- Silica
- Silica
- C8
- C18
- C18
- C8
- Phenyl-Hexyl
- Carbon load:
- 11,7
- 11,7
- 20
- 20
- 11,7
- 15
- 15
- 20
- 15
- 11,7
- 20
- 20
- 11,7
- 15
- USP code:
- L7
- L7
- L1
- L1
- L7
- L11
- L3
- L11
- L1
- L11
- L3
- L3
- L7
- L1
- L1
- L7
- L11
Specifications
Related Information
J.T.Baker® BAKERBOND® Q2100, New Generation UHPLC and HPLC Columns