Orders LinkContactUsLinkComponent
PendoTECH® Single-Use Pressure Sensor Accessories
PendoTECH® Single-Use Pressure Sensor Accessories
PendoTECH® Single-Use Pressure Sensor Accessories
PendoTECH® Single-Use Pressure Sensor Accessories
Optimize performance of your PendoTECH® Pressure Sensor.
  • Choose from a wide range of replacement parts and accessories
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
  • Max. pressure:
  • 76 psi
  • 76 psi
  • 76 psi
  • Process connection:
  • 1/2" x 1/2" hose barb, male luer
  • 1/4" x 1/4" hose barb, male luer
  • 3/8" x 3/8" hose barb, male luer
  • Length:
  • 213,36 cm (84")
  • 304,8 cm (120")
  • 365,76 cm (144")
  • 365,76 cm (144")
  • 182,88 cm (72")
  • 182,88 cm (72")
  • Warranty:
  • 3 Months
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 3 Months
  • 3 Months
  • 2 years
  • 2 years
  • 2 years
  • 1 year
  • 1 year
  • 2 years
  • 2 years
  • 2 years
  • 2 years
Product Family Options
  • 설명
  • Adapter Tee, 1/2" Hose Barb x 1/2" Hose Barb x Male Luer Fitting, Polycarbonate
    1 * 1 items
    1 * 1 items
    CE Compliant
    Specifications
    Cat. No.
    MFLX19406-35
    Max. pressure
    76 psi
    Process connection
    1/2" x 1/2" hose barb, male luer
    Length
    Warranty
    3 Months
    CE Compliant
  • 3-Way Stopcock with Male/Female Luer Inlet /Outlet F Branch, Polysulfone
    1 * 1 items
    1 * 1 items
    Specifications
    Cat. No.
    MFLX19406-49
    Max. pressure
    Process connection
    Length
    Warranty
    2 years
  • Test Cable Assembly for PressureMAT Accuracy Check
    1 * 1 items
    1 * 1 items
    Specifications
    Cat. No.
    MFLX19406-50
    Max. pressure
    Process connection
    Length
    Warranty
    2 years
  • Reusable Temperature Sensor Cable with 1/4" Phone Jack for Luer Sensors; 7 ft (2 m)
    1 * 1 items
    1 * 1 items
    CE Compliant
    Specifications
    Cat. No.
    MFLX19406-14
    Max. pressure
    Process connection
    Length
    213,36 cm (84")
    Warranty
    2 years
    CE Compliant
  • Reusable Temperature Sensor Cable with 1/4" Phone Jack for Hose Barb Fitting; 10 ft (3 m)
    1 * 1 items
    1 * 1 items
    CE Compliant
    Specifications
    Cat. No.
    MFLX19406-15
    Max. pressure
    Process connection
    Length
    304,8 cm (120")
    Warranty
    2 years
    CE Compliant
  • Adapter Tee, 1/4" Hose Barb x 1/4" Hose Barb x Male Luer Fitting, Polycarbonate
    1 * 1 items
    1 * 1 items
    Specifications
    Cat. No.
    MFLX19406-34
    Max. pressure
    76 psi
    Process connection
    1/4" x 1/4" hose barb, male luer
    Length
    Warranty
    3 Months
  • Adapter Tee, 3/8" Hose Barb x 3/8" Hose Barb x Male Luer Fitting, Polycarbonate
    1 * 1 items
    1 * 1 items
    Specifications
    Cat. No.
    MFLX19406-36
    Max. pressure
    76 psi
    Process connection
    3/8" x 3/8" hose barb, male luer
    Length
    Warranty
    3 Months
  • Temperature Sensor Transmitter with 4-20mA Output
    1 * 1 items
    1 * 1 items
    2 year warranty
    Specifications
    Cat. No.
    MFLX19406-45
    Max. pressure
    Process connection
    Length
    Warranty
    2 years
    2 year warranty
  • Temperature Sensor Transmitter DIN Rail Mounting Kit
    1 * 1 items
    1 * 1 items
    2 year warranty
    Specifications
    Cat. No.
    MFLX19406-46
    Max. pressure
    Process connection
    Length
    Warranty
    2 years
    2 year warranty
  • Reusable Temperature Sensor Cable with M8 Termination for Hose Barb Sensors; 12"
    1 * 1 items
    1 * 1 items
    Specifications
    Cat. No.
    MFLX19406-48
    Max. pressure
    Process connection
    Length
    Warranty
    2 years
  • Sensor Cable with 4-wire leads, for 3rd party monitors 12ft (3,7 m)
    1 * 1 items
    1 * 1 items
    Specifications
    Cat. No.
    MFLX19406-80
    Max. pressure
    Process connection
    Length
    365,76 cm (144")
    Warranty
    1 year
  • RS232 Sensor Cable with 4-wire leads, 12 ft (3,7 m)
    1 * 1 items
    1 * 1 items
    Specifications
    Cat. No.
    MFLX19406-82
    Max. pressure
    Process connection
    Length
    365,76 cm (144")
    Warranty
    1 year
  • Cable with RJ12 to Scilog Pump, 6 ft (1,8 m)
    1 * 1 items
    1 * 1 items
    CE Compliant
    Specifications
    Cat. No.
    MFLX19406-85
    Max. pressure
    Process connection
    Length
    182,88 cm (72")
    Warranty
    2 years
    CE Compliant
  • Interface cable 6 ft (1,8 m)
    1 * 1 items
    1 * 1 items
    CE Compliant
    Specifications
    Cat. No.
    MFLX19406-87
    Max. pressure
    Process connection
    Length
    182,88 cm (72")
    Warranty
    2 years
    CE Compliant
  • Benchtop Stand for PressureMAT Monitor
    1 * 1 items
    1 * 1 items
    CE Compliant
    Specifications
    Cat. No.
    MFLX19406-95
    Max. pressure
    Process connection
    Length
    Warranty
    2 years
    CE Compliant
  • Temperature Sensor System Accessory with 4-20mA Output in ABS Plastic Box; 24VDC Wall Supply ***Must order cable separately***
    1 * 1 items
    1 * 1 items
    2 year warranty
    Specifications
    Cat. No.
    MFLX19406-47
    Max. pressure
    Process connection
    Length
    Warranty