Flowline DataView Advanced Meter & Controller
이 제품에 관하여
Easy configuration via push button display module.
- Bright 6-digit LED main display and secondary unit display with relay status indicators
- 1/8 DIN enclosure with NEMA 4X faceplate and shallow depth case
- 32-point linearization function for volumetric tank measurement
- Non-volatile memory with security password protected lock out
- Isolated 4-20 mA repeater output with 40 mA internal power supply
The general purpose meter displays engineering units with one 4-20 mA or 0-10 VDC transmitter input channel and is offered in three configurations with optional 2 or 4 relays, 4 or 8 set points and an isolated 4-20 mA repeater. Each relay can be configured on a single set point or latched on two set points for automatic fill or empty control including duplexing. For field mount installations,
order the single or two meter NEMA box.
3 Options Available Below
Product Details & Documents
Easy configuration via push button display module.
- Bright 6-digit LED main display and secondary unit display with relay status indicators
- 1/8 DIN enclosure with NEMA 4X faceplate and shallow depth case
- 32-point linearization function for volumetric tank measurement
- Non-volatile memory with security password protected lock out
- Isolated 4-20 mA repeater output with 40 mA internal power supply
The general purpose meter displays engineering units with one 4-20 mA or 0-10 VDC transmitter input channel and is offered in three configurations with optional 2 or 4 relays, 4 or 8 set points and an isolated 4-20 mA repeater. Each relay can be configured on a single set point or latched on two set points for automatic fill or empty control including duplexing. For field mount installations,
order the single or two meter NEMA box.
Specifications for Product Family
사양
- Catalog No:
- MFLX23002-18
- MFLX23002-20
- MFLX23002-22
- Signal output:
- 4 to 20 mA
- 4 to 20 mA
- Warranty:
- 1 year
- 1 year
- 1 year
사양
Product Family Options
제품 정보
- 전력재고 여부Price
- MFLX23002-18Advanced Meter, No Outputs; 24 VDC사양:IP65 washdown
More
Specifications
Cat. No.MFLX23002-18Signal outputWarranty1 yearIP65 washdown - MFLX23002-20Advanced Meter/Controller, 2 Relays; 24 VDC사양:IP65 washdown
More
Specifications
Cat. No.MFLX23002-20Signal output4 to 20 mAWarranty1 yearIP65 washdown - MFLX23002-22Advanced Meter/Controller, 4 Relays; 24 VDC사양:IP65 washdown
More
Specifications
Cat. No.MFLX23002-22Signal output4 to 20 mAWarranty1 yearIP65 washdown