Masterflex® Motorized Drum Pump Systems, Avantor®
Complete drum pump system specifically packaged for different applications.
- Flow rate 35 GMP for all models
- Equipped with variable-speed control motors for added safety and control
- Maintenance is reduced with no mechanical seals
- Run dry capability
Pre-configured drum and container pumps are engineered to safely transfer a variety of liquids from drums, tanks, and small storage vessels. Based upon the application, polypropylene (PP), CPVC, PVDF, and 316 stainless steel (SS) materials provide excellent chemical resistance against corrosive liquids.
The Measurement Pump Packages (Models 70614-06, -08 and -09) include a digital flowmeter and totalizer. The dispensing hose for all the pumps is a 6-ft length of PVC tubing. The storage bracket for all packages is made from 16 gauge coated steel.
: Includes electric motor with 16-ft (4.8-m) power cord, pump tube, wall storage bracket, barrel adapter, dispensing hose, and nozzle.
Complete drum pump system specifically packaged for different applications.
- Flow rate 35 GMP for all models
- Equipped with variable-speed control motors for added safety and control
- Maintenance is reduced with no mechanical seals
- Run dry capability
Pre-configured drum and container pumps are engineered to safely transfer a variety of liquids from drums, tanks, and small storage vessels. Based upon the application, polypropylene (PP), CPVC, PVDF, and 316 stainless steel (SS) materials provide excellent chemical resistance against corrosive liquids.
The Measurement Pump Packages (Models 70614-06, -08 and -09) include a digital flowmeter and totalizer. The dispensing hose for all the pumps is a 6-ft length of PVC tubing. The storage bracket for all packages is made from 16 gauge coated steel.
: Includes electric motor with 16-ft (4.8-m) power cord, pump tube, wall storage bracket, barrel adapter, dispensing hose, and nozzle.
- Catalog No:
- MFLX70614-00
- MFLX70614-02
- MFLX70614-03
- MFLX70614-04
- MFLX70614-05
- MFLX70614-06
- MFLX70614-09
- MFLX70614-10
- Model:
- 70614-00
- 70614-02
- 70614-03
- 70614-04
- 70614-05
- 70614-06
- 70614-09
- 70614-10
- Tube length, inlet:
- 39"
- 39"
- 39"
- 39"
- 39"
- 39"
- 39"
- 39"
- Wetted materials:
- CPVC
- Polypropylene
- Polypropylene
- PVDF
- PVDF
- Polypropylene
- PVDF
- Stainless Steel
- Warranty:
- 3 years
- 3 years
- 3 years
- 3 years
- 3 years
- 3 years
- 3 years
- 3 years
Product Family Options
- 설명최대 유량전력
- Motorized Drum Pump Package, 35 gal./min, 39"L CPVC Tube; 115V35 gal./min115 VAC; 60 Hz-
Specifications
Cat. No.MFLX70614-00Model70614-00Tube length, inlet39"Wetted materialsCPVCWarranty3 years - Motorized Drum Pump Package, 35 gal./min, 39"L PP Tube; 115V35 gal./min115 VAC; 50 Hz-
Specifications
Cat. No.MFLX70614-02Model70614-02Tube length, inlet39"Wetted materialsPolypropyleneWarranty3 years - Motorized Drum Pump Package, 35 gal./min, 39"L PP Tube; 230V35 gal./min230 VAC; 60 Hz1 * 1 itemsCE Compliant
Specifications
Cat. No.MFLX70614-03Model70614-03Tube length, inlet39"Wetted materialsPolypropyleneWarranty3 yearsCE Compliant - Motorized Drum Pump Package, 35 gal./min, 39"L PVDF Tube; 115V35 gal./min115 VAC; 60 Hz-
Specifications
Cat. No.MFLX70614-04Model70614-04Tube length, inlet39"Wetted materialsPVDFWarranty3 years - Motorized Drum Pump Package, 35 gal./min, 39"L PVDF Tube; 230V35 gal./min230 VAC; 50 Hz1 * 1 itemsCE Compliant
Specifications
Cat. No.MFLX70614-05Model70614-05Tube length, inlet39"Wetted materialsPVDFWarranty3 yearsCE Compliant - Drum Pump System w/Flow Monitor, 35 gal./min, 39"L PP Tube; 115V35 gal./min115 VAC; 60 Hz-
Specifications
Cat. No.MFLX70614-06Model70614-06Tube length, inlet39"Wetted materialsPolypropyleneWarranty3 years - Drum Pump System w/Flow Monitor, 35 gal./min, 39"L PVDF Tube; 230V35 gal./min230 VAC; 50 Hz1 * 1 itemsCE Compliant
Specifications
Cat. No.MFLX70614-09Model70614-09Tube length, inlet39"Wetted materialsPVDFWarranty3 yearsCE Compliant - Motorized Drum Pump System, 35 gal./min, 39"L SS Tube; 115V35 gal./min115 VAC; 60 Hz-
Specifications
Cat. No.MFLX70614-10Model70614-10Tube length, inlet39"Wetted materialsStainless SteelWarranty3 years