Orders LinkContactUsLinkComponent
John Guest polypropylene threaded elbows
John Guest polypropylene threaded elbows
John Guest polypropylene threaded elbows
Certificates
John Guest polypropylene threaded elbows
4 Options Available Below
Product Details & Documents
Quick, leakproof connection.
  • Polypropylene and EPDM materials meet NSF 51, and NSF 61 requirements for food-grade applications; and is FDA compliant
Push-to-connect fittings offer secure connections and quick disconnection. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semi-rigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing.
Adapters (sold separately) let you permanently modify basic fittings for customized installations.
Specifications for Product Family
사양
  • Max. operating pressure:
  • 150 psi at 70 °F, 60 psi at 140 °F (for water)
  • 150 psi at 70 °F, 60 psi at 140 °F (for water)
  • 150 psi at 70 °F, 60 psi at 140 °F (for water)
  • 150 psi at 70 °F, 60 psi at 140 °F (for water)
  • Max. temperature:
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • Min. temperature:
  • 1 °C (33 °F)
  • 1 °C (33 °F)
  • 1 °C (33 °F)
  • 1 °C (33 °F)
  • Seal material:
  • EPDM
  • EPDM
  • EPDM
  • EPDM
  • Sterilisation:
  • Autoclave only
  • Autoclave only
  • Autoclave only
  • Autoclave only
  • Warranty:
  • 20 years
  • 20 years
  • 20 years
  • 20 years
사양
Product Family Options
제품 정보
  • 연결 유형
    Fitting material
    Flow pattern
    Push-to-connect tube ext.Ø (port 1)
    Thread size NPT(M) (port 2)
    재고 여부
    Price
  • MFLX34007-65
    Threaded Elbow Adapters, 3/8" OD to 3/8" NPT(M), PP, 10/Pk
    Threaded Elbow Adapters, 3/8" OD to 3/8" NPT(M), PP, 10/Pk
    Push-to-Connect to Thread
    Polypropylene
    Elbow
    3/8"
    3/8"
    Log in 재고 확인
    Log in 재고 확인
    1 * 10 items
    1 * 10 items
    사양:
    More
    Specifications
    Cat. No.
    MFLX34007-65
    Max. operating pressure
    150 psi at 70 °F, 60 psi at 140 °F (for water)
    Max. temperature
    60 °C (140 °F)
    Min. temperature
    1 °C (33 °F)
    Seal material
    EPDM
    Sterilisation
    Autoclave only
    Warranty
    20 years
  • MFLX34007-62
    Threaded Elbow Adapters, 1/4" OD to 1/8" NPT(M), PP, 10/Pk
    Threaded Elbow Adapters, 1/4" OD to 1/8" NPT(M), PP, 10/Pk
    Push-to-Connect to Thread
    Polypropylene
    Elbow
    1/4"
    1/8"
    Log in 재고 확인
    Log in 재고 확인
    1 * 10 items
    1 * 10 items
    사양:
    More
    Specifications
    Cat. No.
    MFLX34007-62
    Max. operating pressure
    150 psi at 70 °F, 60 psi at 140 °F (for water)
    Max. temperature
    60 °C (140 °F)
    Min. temperature
    1 °C (33 °F)
    Seal material
    EPDM
    Sterilisation
    Autoclave only
    Warranty
    20 years
  • MFLX34007-63
    Threaded Elbow Adapters, 1/4" OD to 1/4" NPT(M), PP, 10/Pk
    Threaded Elbow Adapters, 1/4" OD to 1/4" NPT(M), PP, 10/Pk
    Push-to-Connect to Thread
    Polypropylene
    Elbow
    1/4"
    1/4"
    Log in 재고 확인
    Log in 재고 확인
    1 * 10 items
    1 * 10 items
    사양:
    More
    Specifications
    Cat. No.
    MFLX34007-63
    Max. operating pressure
    150 psi at 70 °F, 60 psi at 140 °F (for water)
    Max. temperature
    60 °C (140 °F)
    Min. temperature
    1 °C (33 °F)
    Seal material
    EPDM
    Sterilisation
    Autoclave only
    Warranty
    20 years
  • MFLX34007-64
    Threaded Elbow Adapters, 3/8" OD to 1/4" NPT(M), PP, 10/Pk
    Threaded Elbow Adapters, 3/8" OD to 1/4" NPT(M), PP, 10/Pk
    Push-to-Connect to Thread
    Polypropylene
    Elbow
    3/8"
    1/4"
    Log in 재고 확인
    Log in 재고 확인
    1 * 10 items
    1 * 10 items
    사양:
    More
    Specifications
    Cat. No.
    MFLX34007-64
    Max. operating pressure
    150 psi at 70 °F, 60 psi at 140 °F (for water)
    Max. temperature
    60 °C (140 °F)
    Min. temperature
    1 °C (33 °F)
    Seal material
    EPDM
    Sterilisation
    Autoclave only
    Warranty
    20 years
Accessories
제품 정보
  • 제품 번호
    연결 유형
    Fitting material
    재고 여부
    Price
  • Push-To-Connect Fitting, Male Thread, Polypropylene, 1/4" OD×1/8" NPT(M); 10/Pk
    Push-To-Connect Fitting, Male Thread, Polypropylene, 1/4" OD×1/8" NPT(M); 10/Pk
    MFLX34007-53
    Push-to-Connect to Thread
    Polypropylene
    Log in 재고 확인
    1 * 10 items
제품 정보
  • 제품 번호
    연결 유형
    Fitting material
    재고 여부
    Price
  • Push-To-Connect Fitting, Male Thread, Polypropylene, 1/4" OD×1/4" NPT(M); 10/Pk
    Push-To-Connect Fitting, Male Thread, Polypropylene, 1/4" OD×1/4" NPT(M); 10/Pk
    MFLX34007-54
    Push-to-Connect to Thread
    Polypropylene
    Log in 재고 확인
    1 * 10 items
제품 정보
  • 제품 번호
    연결 유형
    Fitting material
    재고 여부
    Price
  • Push-To-Connect Fitting, Male Thread, Polypropylene, 3/8" OD×1/4" NPT(M); 10/Pk
    Push-To-Connect Fitting, Male Thread, Polypropylene, 3/8" OD×1/4" NPT(M); 10/Pk
    MFLX34007-55
    Push-to-Connect to Thread
    Polypropylene
    Log in 재고 확인
    1 * 10 items
제품 정보
  • 제품 번호
    연결 유형
    Fitting material
    재고 여부
    Price
  • Push-To-Connect Fitting, Male Thread, Polypropylene, 3/8" OD×3/8" NPT(M); 10/Pk
    Push-To-Connect Fitting, Male Thread, Polypropylene, 3/8" OD×3/8" NPT(M); 10/Pk
    MFLX34007-56
    Push-to-Connect to Thread
    Polypropylene
    Log in 재고 확인
    1 * 10 items
제품 정보
  • 제품 번호
    연결 유형
    Fitting material
    재고 여부
    Price
  • Push-to-connect reducer adapters, polypropylene, 3/8"×1/4", 10/pack
    Push-to-connect reducer adapters, polypropylene, 3/8"×1/4", 10/pack
    MFLX34007-57
    Push-to-Connect Reducer
    Polypropylene
    Log in 재고 확인
    1 * 1 items
제품 정보
  • 제품 번호
    연결 유형
    Fitting material
    재고 여부
    Price
  • Push-To-Connect Fitting, Elbow Adapter, Polypropylene, ¼" OD×¼" OD; 10/Pk
    Push-To-Connect Fitting, Elbow Adapter, Polypropylene, ¼" OD×¼" OD; 10/Pk
    MFLX34007-58
    Push-to-Connect Union
    Polypropylene
    Log in 재고 확인
    1 * 10 items
제품 정보
  • 제품 번호
    연결 유형
    Fitting material
    재고 여부
    Price
  • Push-To-Connect Fitting, Elbow Adapter, Polypropylene, ⅜" OD×⅜" OD; 10/Pk
    Push-To-Connect Fitting, Elbow Adapter, Polypropylene, ⅜" OD×⅜" OD; 10/Pk
    MFLX34007-60
    Push-to-Connect Union
    Polypropylene
    Log in 재고 확인
    1 * 10 items