Masterflex® I/P® Pump Head Adapters, Avantor®
About this item
Connect any I/P® pump head to a 56C frame motor.
- Cast aluminum adapters feature built-in gear box for easy setup
- Compatible with all I/P pump heads
- Suitable for use with 1⁄10 hp to 1⁄2 hp 56C frame motor and 650 rpm max pump head speed
These I/P® pump head adapters work with 56C frame motors, allowing you to customize a pump for your application. Add a motor controller for variable-speed and/or remote-control operation. Adapters are compatible with all I/P pump heads.
: Includes adapter with built-in gear box, hardware.
4 Options Available Below
Product Details & Documents
Connect any I/P® pump head to a 56C frame motor.
- Cast aluminum adapters feature built-in gear box for easy setup
- Compatible with all I/P pump heads
- Suitable for use with 1⁄10 hp to 1⁄2 hp 56C frame motor and 650 rpm max pump head speed
These I/P® pump head adapters work with 56C frame motors, allowing you to customize a pump for your application. Add a motor controller for variable-speed and/or remote-control operation. Adapters are compatible with all I/P pump heads.
: Includes adapter with built-in gear box, hardware.
Specifications for Product Family
Specifications
- Model:
- 77490-00
- 77490-10
- 77490-20
- 77490-30
- Drive height:
- 165 mm (6,5")
- 165 mm (6,5")
- 165 mm (6,5")
- 165 mm (6,5")
- Drive length:
- 152 mm (6")
- 152 mm (6")
- 171 mm (6,75")
- 171 mm (6,75")
- Drive width:
- 165 mm (6,5")
- 165 mm (6,5")
- 165 mm (6,5")
- 165 mm (6,5")
- Gear ratio:
- 3,1:1
- 4,8:1
- 9,7:1
- 17,8:1
- Min. RPM:
- 20 rpm
- 20 rpm
- 20 rpm
- 20 rpm
- Motor mount:
- NEMA Type 56C
- NEMA Type 56C
- NEMA Type 56C
- NEMA Type 56C
- Motor requirements:
- 1/4 hp to 1/2 hp; 2015 max motor rpm
- 1/4 hp to 1/2 hp; 3120 max motor rpm
- 1/4 hp to 1/2 hp; 3600 max motor rpm
- 1/4 hp to 1/2 hp; 4800 max motor rpm
- Mounting type:
- Direct coupling
- Direct coupling
- Direct coupling
- Direct coupling
- Open-head sensor:
- No
- No
- No
- No
- Pump series:
- I/P
- I/P
- I/P
- I/P
- Warranty:
- 2 years
- 2 years
- 2 years
- 2 years
Specifications
Product Family Options
Product Information
- 설명No. of pump heads accepted최대 유량Max. RPMHousing materialAvailabilityPrice
- I/P NEMA Type 56 C Pump Head Adapter, 650 rpm max, 3,1:1 gear ratio217000 ml/min650 rpmPainted aluminiumLog in for availability1 * 1 itemsSpecifications:2 year warranty
More
Specifications
Cat. No.MFLX77490-00Model77490-00Drive height165 mm (6,5")Drive length152 mm (6")Drive width165 mm (6,5")Gear ratio3,1:1Min. RPM20 rpmMotor mountNEMA Type 56CMotor requirements1/4 hp to 1/2 hp; 2015 max motor rpmMounting typeDirect couplingOpen-head sensorNoPump seriesI/PWarranty2 years2 year warranty - I/P NEMA Type 56 C Pump Head Adapter, 650 rpm max, 4,8:1 gear ratio217000 ml/min650 rpmPainted aluminiumLog in for availability1 * 1 itemsSpecifications:2 year warranty
More
Specifications
Cat. No.MFLX77490-10Model77490-10Drive height165 mm (6,5")Drive length152 mm (6")Drive width165 mm (6,5")Gear ratio4,8:1Min. RPM20 rpmMotor mountNEMA Type 56CMotor requirements1/4 hp to 1/2 hp; 3120 max motor rpmMounting typeDirect couplingOpen-head sensorNoPump seriesI/PWarranty2 years2 year warranty - I/P NEMA Type 56 C Pump Head Adapter, 650 rpm max, 9,7:1 gear ratio29300 ml/min356 rpmPainted aluminiumLog in for availability1 * 1 itemsSpecifications:2 year warranty
More
Specifications
Cat. No.MFLX77490-20Model77490-20Drive height165 mm (6,5")Drive length171 mm (6,75")Drive width165 mm (6,5")Gear ratio9,7:1Min. RPM20 rpmMotor mountNEMA Type 56CMotor requirements1/4 hp to 1/2 hp; 3600 max motor rpmMounting typeDirect couplingOpen-head sensorNoPump seriesI/PWarranty2 years2 year warranty - I/P NEMA Type 56 C pump head adapter, 17,8:1 adapter gear ratio25000 ml/min194 rpmPainted aluminiumLog in for availability1 * 1 itemsSpecifications:2 year warranty
More
Specifications
Cat. No.MFLX77490-30Model77490-30Drive height165 mm (6,5")Drive length171 mm (6,75")Drive width165 mm (6,5")Gear ratio17,8:1Min. RPM20 rpmMotor mountNEMA Type 56CMotor requirements1/4 hp to 1/2 hp; 4800 max motor rpmMounting typeDirect couplingOpen-head sensorNoPump seriesI/PWarranty2 years2 year warranty