Masterflex® Plastic Assorted Fitting Kits, Avantor®
Get organized today!
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Assorted fittings come in an impact-resistant plastic box with secure snap-shut latches to prevent spillage
- Labeled compartments let you easily locate the fittings you want
- Order storage racks separately
Organize your workstation with these convenient fitting kits. Assorted fittings come in impact resistant plastic box with secure latch to keep fittings organized and in place. Label compartments make fitting location quick and easy. Choose from four materials: high-density polyethylene (HDPE), black nylon, natural polypropylene, and natural PVDF.
Get organized today!
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Assorted fittings come in an impact-resistant plastic box with secure snap-shut latches to prevent spillage
- Labeled compartments let you easily locate the fittings you want
- Order storage racks separately
Organize your workstation with these convenient fitting kits. Assorted fittings come in impact resistant plastic box with secure latch to keep fittings organized and in place. Label compartments make fitting location quick and easy. Choose from four materials: high-density polyethylene (HDPE), black nylon, natural polypropylene, and natural PVDF.
- Catalog No:
- MFLX31514-01
- MFLX31514-02
- MFLX31514-05
- MFLX31514-06
- MFLX31514-09
- MFLX31514-10
- MFLX31514-13
- MFLX31514-14
- MFLX31514-17
- MFLX31514-18
- MFLX31514-22
- MFLX31514-25
- MFLX31514-26
- MFLX31514-29
- MFLX31514-32
- MFLX31514-33
- MFLX31514-34
- MFLX41514-03
- MFLX41514-07
- MFLX41514-11
- MFLX41514-15
- MFLX41514-19
- MFLX41514-23
- MFLX41514-27
- MFLX41514-30
- MFLX41514-35
- Model:
- 31514-01
- 31514-02
- 31514-05
- 31514-06
- 31514-09
- 31514-10
- 31514-13
- 31514-14
- 31514-17
- 31514-18
- 31514-22
- 31514-25
- 31514-26
- 31514-29
- 31514-32
- 31514-33
- 31514-34
- 41514-03
- 41514-07
- 41514-11
- 41514-15
- 41514-19
- 41514-23
- 41514-27
- 41514-30
- 41514-35
- Max. temperature:
- 87 °C (190 °F)
- 93 °C (200 °F)
- 87 °C (190 °F)
- 93 °C (200 °F)
- 87 °C (190 °F)
- 93 °C (200 °F)
- 87 °C (190 °F)
- 93 °C (200 °F)
- 87 °C (190 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 87 °C (190 °F)
- 93 °C (200 °F)
- 93 °C (200 °F)
- 87 °C (190 °F)
- 87 °C (190 °F)
- 93 °C (200 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- 135 °C (275 °F)
- Min. temperature:
- –53 °C (–65 °F)
- –40 °C (–40 °F)
- –53 °C (–65 °F)
- –40 °C (–40 °F)
- –53 °C (–65 °F)
- –40 °C (–40 °F)
- –53 °C (–65 °F)
- –40 °C (–40 °F)
- –53 °C (–65 °F)
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- –53 °C (–65 °F)
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- –53 °C (–65 °F)
- –53 °C (–65 °F)
- –40 °C (–40 °F)
- –17 °C (0 °F)
- –17 °C (0 °F)
- –17 °C (0 °F)
- –17 °C (0 °F)
- –17 °C (0 °F)
- –17 °C (0 °F)
- –17 °C (0 °F)
- –17 °C (0 °F)
- –17 °C (0 °F)
- Warranty:
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 3 Months
- 3 Months
- 3 Months
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 1 Month
- 3 Months
Product Family Options
- 설명연결 유형Fitting materialFlow pattern
- Fitting Kit, HDPE, Straight/Reducer/Plug, Hose Barb, Small FittingsKitHDPEStraight1 * 1 itemsCE certificate
Specifications
Cat. No.MFLX31514-01Model31514-01Max. temperature87 °C (190 °F)Min. temperature–53 °C (–65 °F)Warranty1 Month - Fitting Kit, Black Nylon, Straight/Reducer/Plug, Hosebarb, Small FittingsKitBlack NylonStraight1 * 1 items
Specifications
Cat. No.MFLX31514-02Model31514-02Max. temperature93 °C (200 °F)Min. temperature–40 °C (–40 °F)Warranty1 Month - Fitting Kit, HDPE, Straight and Reducer, Hose Barb, Large FittingsKitHDPEStraight1 * 1 itemsCE certificate
Specifications
Cat. No.MFLX31514-05Model31514-05Max. temperature87 °C (190 °F)Min. temperature–53 °C (–65 °F)Warranty1 Month - Fitting Kit, Black Nylon, Straight and Reducer, Hose Barb, Large Fittings; 1 SetKitNylonStraight1 * 1 items
Specifications
Cat. No.MFLX31514-06Model31514-06Max. temperature93 °C (200 °F)Min. temperature–40 °C (–40 °F)Warranty1 Month - Fitting Kit, HDPE, Straight and Reducer, Threaded Adapter to Hose Barb; 1 SetKitHDPEStraight1 * 1 itemsCE certificate
Specifications
Cat. No.MFLX31514-09Model31514-09Max. temperature87 °C (190 °F)Min. temperature–53 °C (–65 °F)Warranty1 Month - Fitting Kit, Black Nylon, Straight and Reducer, Thread Adapter to HosebarbKitNylonStraight1 * 1 items
Specifications
Cat. No.MFLX31514-10Model31514-10Max. temperature93 °C (200 °F)Min. temperature–40 °C (–40 °F)Warranty1 Month - CE certificate
Specifications
Cat. No.MFLX31514-13Model31514-13Max. temperature87 °C (190 °F)Min. temperature–53 °C (–65 °F)Warranty1 Month Specifications
Cat. No.MFLX31514-14Model31514-14Max. temperature93 °C (200 °F)Min. temperature–40 °C (–40 °F)Warranty1 Month- CE certificate
Specifications
Cat. No.MFLX31514-17Model31514-17Max. temperature87 °C (190 °F)Min. temperature–53 °C (–65 °F)Warranty1 Month Specifications
Cat. No.MFLX31514-18Model31514-18Max. temperature93 °C (200 °F)Min. temperature–40 °C (–40 °F)Warranty1 Month- Fitting Kit, Black Nylon, Elbow/Tee/Nipple/Plug, Thread Adapter to HosebarbKitBlack NylonElbow1 * 1 items
Specifications
Cat. No.MFLX31514-22Model31514-22Max. temperature93 °C (200 °F)Min. temperature–40 °C (–40 °F)Warranty1 Month - Fitting Kit, HDPE, Elbow and Tee, Threaded Adapter to Hose Barb; 1 SetKitHDPET1 * 1 itemsCE certificate
Specifications
Cat. No.MFLX31514-25Model31514-25Max. temperature87 °C (190 °F)Min. temperature–53 °C (–65 °F)Warranty1 Month - Fitting Kit, Black Nylon, Elbow and Tee, Threaded Adapter to Hose Barb; 1 SetKitBlack NylonT1 * 1 items
Specifications
Cat. No.MFLX31514-26Model31514-26Max. temperature93 °C (200 °F)Min. temperature–40 °C (–40 °F)Warranty1 Month - Fitting Kit, Black Nylon, Straight/Reducer/Elbow/Tee, BSP Thread to HosebarbKitBlack NylonT1 * 1 items
Specifications
Cat. No.MFLX31514-29Model31514-29Max. temperature93 °C (200 °F)Min. temperature–40 °C (–40 °F)Warranty1 Month - Fitting Kit, HDPE, Straight/Reducer/Elbow/Tee, BSP Threaded Adapter to HosebarbKitHDPET1 * 1 itemsCE certificate
Specifications
Cat. No.MFLX31514-32Model31514-32Max. temperature87 °C (190 °F)Min. temperature–53 °C (–65 °F)Warranty3 Months - Fitting Kit, HDPE, Straight and Reducer, Threaded Adapter to Hose Barb and Threaded Adapter Union; 1 SetKitHDPEStraight1 * 1 itemsCE certificate
Specifications
Cat. No.MFLX31514-33Model31514-33Max. temperature87 °C (190 °F)Min. temperature–53 °C (–65 °F)Warranty3 Months - Fitting Kit, Black Nylon, Straight and Reducer, Threaded Adapter to Hose Barb and Threaded Adapter Union; 1 SetKitBlack NylonStraight1 * 1 items
Specifications
Cat. No.MFLX31514-34Model31514-34Max. temperature93 °C (200 °F)Min. temperature–40 °C (–40 °F)Warranty3 Months - Fitting Kit, Polypropylene, Straight/Reducer/Plug, Hosebarb, Small FittingsKitPolypropyleneStraight1 * 1 itemsCE certificate
Specifications
Cat. No.MFLX41514-03Model41514-03Max. temperature135 °C (275 °F)Min. temperature–17 °C (0 °F)Warranty1 Month - Fitting Kit, Polypropylene, Straight and Reducer, Hosebarb, Large FittingsKitPolypropyleneStraight1 * 1 itemsCE certificate
Specifications
Cat. No.MFLX41514-07Model41514-07Max. temperature135 °C (275 °F)Min. temperature–17 °C (0 °F)Warranty1 Month - Fitting Kit, Polypropylene, Straight and Reducer, Threaded Adapter to HosebarbKitPolypropyleneStraight1 * 1 itemsCE certificate
Specifications
Cat. No.MFLX41514-11Model41514-11Max. temperature135 °C (275 °F)Min. temperature–17 °C (0 °F)Warranty1 Month - Fitting Kit, Polypropylene, Elbow/Tee/Y, Hose Barb; 1 SetKitPolypropyleneElbow1 * 1 itemsCE certificate
Specifications
Cat. No.MFLX41514-15Model41514-15Max. temperature135 °C (275 °F)Min. temperature–17 °C (0 °F)Warranty1 Month - CE certificate
Specifications
Cat. No.MFLX41514-19Model41514-19Max. temperature135 °C (275 °F)Min. temperature–17 °C (0 °F)Warranty1 Month - Fitting Kit, Polypropylene, Elbow/Tee/Nipple/Plug, Threaded Adapter to HosebarbKitPolypropyleneElbow1 * 1 itemsCE certificate
Specifications
Cat. No.MFLX41514-23Model41514-23Max. temperature135 °C (275 °F)Min. temperature–17 °C (0 °F)Warranty1 Month - Fitting Kit, Polypropylene, Elbow and Tee, Threaded Adapter to Hose Barb; 1 SetKitPolypropyleneT1 * 1 itemsCE certificate
Specifications
Cat. No.MFLX41514-27Model41514-27Max. temperature135 °C (275 °F)Min. temperature–17 °C (0 °F)Warranty1 Month - Fitting Kit, Polypropylene, Straight/Reducer/Elbow/Tee, BSP Thread to HosebarbKitPolypropyleneT1 * 1 itemsCE certificate
Specifications
Cat. No.MFLX41514-30Model41514-30Max. temperature135 °C (275 °F)Min. temperature–17 °C (0 °F)Warranty1 Month - Fitting Kit, Polypropylene, Straight and Reducer, Threaded Adapter to Hose Barb and Threaded Adapter Union; 1 SetKitPolypropyleneStraight1 * 1 itemsCE certificate
Specifications
Cat. No.MFLX41514-35Model41514-35Max. temperature135 °C (275 °F)Min. temperature–17 °C (0 °F)Warranty3 Months