Masterflex® Ismatec® ISM738 Plate for Mounting Masterflex® Pump Heads to Quick-Couple Drives
Catalog # MFLX78020-90
Supplier: Avantor Fluid Handling
Specifications
- 설명:Mounting Plate for Heads to QuickCouple Drive. 1 year warranty
- Pk:1
Specifications
About this item
Adapt your drive to accept Masterflex® pump heads.
- Mounting plate attaches quickly and easily
This is the mounting plate needed to attach a Masterflex® pump head to an Ismatec® BVP / MCP Quick-Couple Drive.
- 설명:Mounting Plate for Heads to QuickCouple Drive. 1 year warranty
- Environmentally Preferable:
- Pk:1