Orders LinkContactUsLinkComponent
VWR®, Pipette Controller, Classic
VWR®, Pipette Controller, Classic
공급사:   VWR Collection
VWR®, Pipette Controller, Classic
공급사:  VWR Collection
Certificates
VWR®, Pipette Controller, Classic
VWR®, Pipette Controller, Classic
4 Options Available Below
Product Details & Documents
Pipette controller for glass and plastic pipettes from 1 to 100 ml.
  • Speed of suction and dispensing controlled through the finger triggers
  • Fully charged has approximately 8 h of continuous use available
  • Low battery light illuminates when 1 hour's power remains
Offers a unique combination of features to give precision fingertip control in a classic streamlined design, suitable for users with large or small hands. Gravity dispense mode is designed for use with 'To Deliver' (TD) pipettes.
Product Family Options
제품 정보
  • Colour code
    재고 여부
    Price
  • Blue
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
  • Green
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
  • Orange
    Log in 재고 확인
    Log in 재고 확인
    1 * 1 items
    1 * 1 items
  • Red