Orders LinkContactUsLinkComponent
VWR 실험용 니트릴 장갑
VWR 실험용 니트릴 장갑
VWR 실험용 니트릴 장갑
VWR 실험용 니트릴 장갑
이 장갑은 합성 니트릴로 만들어졌으며 높은 내화학성과 내구성으로 다양한 실험에 적합합니다. 이 장갑은 미생물 및 바이러스 연구를 위한 EN ISO 374-5 표준을 충족합니다.
  • 파우더프리, 비멸균, 일회용
  • 우수한 그립감을 위해 손 끝 돌기처리
  • 롤림(rolled rim)으로 소매 미끄러짐 방지
  • 양손잡이형
  • 박테리오파지 침투 테스트 포함 EN ISO 374-5 인증
  • AQL 1.5를 만족하는 고품질 제품
VWR® 실험용 니트릴 장갑은 0.1mm 두께를 가지지만 최고의 안전 수준을 가집니다. 이 장갑은 라텍스 알러지를 가진 연구원들을 위한 최적의 대안입니다.
 : ISO 9001, ISO 13485, ISO 14001, Medical Device Regulation (EU) 2014-746: Class 1, PPE Regulation (EU) 2016-425: Category I or III, Regulation (EC) 1935/2004 on Food Contact Materials. EN 420, EN ISO 374-1, EN 374-2, EN 16523-1, EN 374-4, EN ISO 374-5 (subject to labelling), EN 455-1, EN 455-2, EN 455-3, EN 455-4, ISO 2859-1, ASTM D6319, ASTM F1671.
  • Catalog No:
  • 112-6090
  • 112-6091
  • 112-6092
  • 112-6093
  • 112-6094
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Material:
  • Nitrile
  • Nitrile
  • Nitrile
  • Nitrile
  • Nitrile
  • Finish:
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Textured fingertips
  • Cuff style:
  • Rolled
  • Rolled
  • Rolled
  • Rolled
  • Rolled
Product Family Options
  • 장갑 사이즈
    Colour
    길이
    두께
  • XS
    Blue
    240 mm
    0,10 mm
    1 * 100 items
    1 * 100 items - ₩7,003
    ₩7,003
    1 * 100 items
       ₩7,003
    Specifications
    Cat. No.
    112-6090
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Material
    Nitrile
    Finish
    Textured fingertips
    Cuff style
    Rolled
  • S
    Blue
    240 mm
    0,10 mm
    1 * 100 items
    1 * 100 items - ₩7,003
    ₩7,003
    1 * 100 items
       ₩7,003
    Specifications
    Cat. No.
    112-6091
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Material
    Nitrile
    Finish
    Textured fingertips
    Cuff style
    Rolled
  • M
    Blue
    240 mm
    0,10 mm
    1 * 100 items
    1 * 100 items - ₩7,003
    ₩7,003
    1 * 100 items
       ₩7,003
    Specifications
    Cat. No.
    112-6092
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Material
    Nitrile
    Finish
    Textured fingertips
    Cuff style
    Rolled
  • L
    Blue
    240 mm
    0,10 mm
    1 * 100 items
    1 * 100 items - ₩7,003
    ₩7,003
    1 * 100 items
       ₩7,003
    Specifications
    Cat. No.
    112-6093
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Material
    Nitrile
    Finish
    Textured fingertips
    Cuff style
    Rolled
  • XL
    Blue
    240 mm
    0,10 mm
    1 * 90 items
    1 * 90 items - ₩7,003
    ₩7,003
    1 * 90 items
       ₩7,003
    Specifications
    Cat. No.
    112-6094
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex-free
    Powdered / powder-free
    Powder-free
    Material
    Nitrile
    Finish
    Textured fingertips
    Cuff style
    Rolled