ContactUsLinkComponent
VWR 실험용 라텍스 장갑
undefined
VWR 실험용 라텍스 장갑
  • Catalog No:
  • 112-6087
  • 112-6086
  • 112-6089
  • 112-6085
  • 112-6088
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex
  • Latex
  • Latex
  • Latex
  • Latex
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Material:
  • Natural latex
  • Natural latex
  • Natural latex
  • Natural latex
  • Natural latex
  • Finish:
  • Grippy, finger textured
  • Grippy, finger textured
  • Grippy, finger textured
  • Grippy, finger textured
  • Grippy, finger textured
  • Cuff style:
  • Rolled
  • Rolled
  • Rolled
  • Rolled
  • Rolled
  • EN norm:
  • EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
  • EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
  • EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
  • EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
  • EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
Frequently Bought Together