Orders LinkContactUsLinkComponent
VWR 실험용 라텍스 장갑
VWR 실험용 라텍스 장갑
공급사:   VWR Collection
VWR 실험용 라텍스 장갑
공급사:  VWR Collection
Certificates
VWR 실험용 라텍스 장갑
5 Options Available Below
Product Details & Documents
VWR 라텍스 장갑은 화학물질 취급을 위한 EN 374 산업표준과 함께 유럽 의료기기 인증인 EN 455 표준을 충족합니다.
  • 파우더프리, 비멸균, 일회용
  • 우수한 그립감을 위해 손 끝 돌기처리
  • 롤림(rolled rim)으로 소매 미끄러짐 방지
  • 양손잡이형
  • AQL 1.5를 만족하는 고품질 제품
특수 표면 처리 기술로 손쉬운 착용 가능, 천연 라텍스 소재로 장시간 사용에도 높은 편안함 제공
Certifications: EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS; Medical Device Regulation (EU) 2017/745: Class 1
Food contact approved
Specifications for Product Family
사양
  • Catalog No:
  • 112-6085
  • 112-6086
  • 112-6087
  • 112-6088
  • 112-6089
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex / latex-free:
  • Latex
  • Latex
  • Latex
  • Latex
  • Latex
  • Powdered / powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Material:
  • Natural latex
  • Natural latex
  • Natural latex
  • Natural latex
  • Natural latex
  • Finish:
  • Grippy, finger textured
  • Grippy, finger textured
  • Grippy, finger textured
  • Grippy, finger textured
  • Grippy, finger textured
  • Cuff style:
  • Rolled
  • Rolled
  • Rolled
  • Rolled
  • Rolled
  • EN norm:
  • EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
  • EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
  • EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
  • EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
  • EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
사양
Product Family Options
제품 정보
  • 장갑 사이즈
    Colour
    길이
    두께
    Packed
    재고 여부
    Price
  • XS
    White
    240 mm
    0,21 mm
    100/dispenser
    Log in 재고 확인
    Log in 재고 확인
    1 * 100 items
    1 * 100 items
    사양:
    More
    Specifications
    Cat. No.
    112-6085
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex
    Powdered / powder-free
    Powder-free
    Material
    Natural latex
    Finish
    Grippy, finger textured
    Cuff style
    Rolled
    EN norm
    EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
  • S
    White
    240 mm
    0,21 mm
    100/dispenser
    Log in 재고 확인
    Log in 재고 확인
    1 * 100 items
    1 * 100 items
    사양:
    More
    Specifications
    Cat. No.
    112-6086
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex
    Powdered / powder-free
    Powder-free
    Material
    Natural latex
    Finish
    Grippy, finger textured
    Cuff style
    Rolled
    EN norm
    EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
  • M
    White
    240 mm
    0,21 mm
    100/dispenser
    Log in 재고 확인
    Log in 재고 확인
    1 * 100 items
    1 * 100 items
    사양:
    More
    Specifications
    Cat. No.
    112-6087
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex
    Powdered / powder-free
    Powder-free
    Material
    Natural latex
    Finish
    Grippy, finger textured
    Cuff style
    Rolled
    EN norm
    EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
  • L
    White
    240 mm
    0,21 mm
    100/dispenser
    Log in 재고 확인
    Log in 재고 확인
    1 * 100 items
    1 * 100 items
    사양:
    More
    Specifications
    Cat. No.
    112-6088
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex
    Powdered / powder-free
    Powder-free
    Material
    Natural latex
    Finish
    Grippy, finger textured
    Cuff style
    Rolled
    EN norm
    EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS
  • XL
    White
    240 mm
    0,21 mm
    90/dispenser
    Log in 재고 확인
    Log in 재고 확인
    1 * 90 items
    1 * 90 items
    사양:
    More
    Specifications
    Cat. No.
    112-6089
    Ambidextrous / hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex / latex-free
    Latex
    Powdered / powder-free
    Powder-free
    Material
    Natural latex
    Finish
    Grippy, finger textured
    Cuff style
    Rolled
    EN norm
    EN 455, EN ISO 374-1 Type B (KPT), EN ISO 374-5 VIRUS