Avantor® ACE® UltraCore C18, Analytical HPLC/UHPLC Columns
CORE-35F-1030
- 컬럼 형식:Analytical
- 내부 직경:3,00 mm
- 입자 크기:3,5 µm
- 기공 크기:95 Å
- Max. pressure:600 bar
- 길이:100 mm
- Seq:05
- Endcapped:Yes
- USP code:L1
- pH range:2 - 9
- Surface area:115 m²/g
- Temperature limit:60 °C (pH 2); 40 °C (pH 9)
- Carbon load:8,2%
- Column hardware:Stainless steel
- Phase:C18
- Pk:1
Avantor® ACE® UltraCore C18 are high performing solid-core (core-shell) particle columns designed for a wide range of LC applications. This densely bonded octadecyl phase can be used to separate acidic, basic and neutral molecules, based on hydrophobic retention mechanisms.
- High purity, base deactivated silica with excellent reproducibility
- High efficiency separations comparable with sub-2 µm packings but lower back pressure
- Ideal for a wide range of analytes differing in hydrophobicity
These stainless steel columns are available in a variety of dimensions and are highly compatible with LC-MS and high sensitivity applications.