VWR®, Pesticide reference standard, Performance Check Solution pesticides and PCBs, EPA 608
제품 번호 124802M
공급사: VWR Chemicals
CAS 번호:
사양
- 포장 유형:Vial
- Pk:1 mL
- 설명:Performance Check Solution pesticides and PCBs, EPA 608
사양
이 제품에 관하여
17 components.
20 µg/ml : Aldrin; a-BHC; b-BHC; d-BHC; g-BHC; p,p'-DDD; p,p'-DDE; Dieldrin; Endosulfan I; Endrin aldehyde; Heptachlor; Heptachlor epoxide (Isomer B); Methoxychlor
100 µg/ml : p,p'-DDT; Endosulfan II; Endosulfan sulfate; Endrin