Orders LinkContactUsLinkComponent
Ninhydrin reagent kit (for buffer PF)
Ninhydrin reagent kit (for buffer PF)
1
Catalog # HPLCNINKIT.
Supplier:   VWR Chemicals
Ninhydrin reagent kit (for buffer PF)
Catalog # HPLCNINKIT.
Supplier:  VWR Chemicals
CAS Number:  
Ninhydrin reagent kit (for buffer PF)
Ninhydrin reagent kit (for buffer PF)
1
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
  • EINECS:000-000-0
Specifications
Specifications
  • Pk:
    1
Specifications