Avantor® ACE® Generix® C18 & C18(2), Analytical HPLC Columns
Supplier: Avantor
Certificates
About this item
These high performance HPLC columns contain high purity spherical silica.
- Ultra-narrow particle size distribution for excellent column efficiency
- Sharp, highly symmetrical peaks for acidic, basic and neutral analytes
- Excellent column to column and batch to batch reproducibility
Each column is individually tested and supplied with a test chromatogram.
Two generations of Avantor® ACE® Generix® C18 exist: C18 and C18(2) they differ in pore size, carbon load and surface area.
Specifications
- Catalog No:
- GEN-3C182-1202
- GEN-3C182-1002
- GEN-3C182-7502
- GEN-3C182-0502
- GEN-3C182-1546
- GEN-3C182-1046
- GEN-3C182-1502
- ACETGEN-3C18-0502
- GEN-5C18-1546
- GEN-3C18-GD
- GEN5C1820502/3
- GEN-3C182-1204
- GEN-3C182-1004
- GEN3C1821546/3
- GEN3C1821046/3
- GEN-3C182-7504
- GEN-3C182-0504
- GEN3C1820502/3
- GEN-3C182-1503
- GEN-3C182-1203
- GEN-3C182-1003
- GEN-3C182-7503
- GEN-3C182-0503
- GEN-3C182-0546
- GEN-3C182-1504
- GEN-5C182-2502
- GEN-5C182-1502
- GEN-5C182-1202
- GEN5C1822546/3
- GEN-5C182-1002
- GEN5C1821546/3
- GEN-5C182-7502
- GEN-5C182-0502
- GEN-3C182-1246
- GEN-3C182-7546
- GEN-5C182-1003
- GEN-5C182-7503
- GEN-5C182-0503
- GEN-5C182-1546
- GEN-5C182-2546
- GEN-5C18-GD
- GEN-5C18-2546
- GEN-5C182-0504
- GEN-5C182-2503
- GEN-5C182-1503
- GEN-5C182-1203
- GEN-C182-GD
- GEN-C182-GD2
- ACETGEN-5C18-1046
- ACETGEN-3C18-1546
- ACETGEN-3C18-1046
- ACETGEN-3C18-1502
- ACETGEN-3C18-1002
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Phase name:
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18
- C18
- C18
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18
- C18
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18(2)
- C18
- C18
- C18
- C18
- C18
- Carbon load:
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 15,0%
- 15,0%
- 15,0%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 15,0%
- 15,0%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 20,3%
- 15,0%
- 15,0%
- 15,0%
- 15,0%
- 15,0%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- Surface area:
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 280 m²/g
- 280 m²/g
- 280 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 280 m²/g
- 280 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 280 m²/g
- 280 m²/g
- 280 m²/g
- 280 m²/g
- 280 m²/g
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)