Orders LinkContactUsLinkComponent
Rankem®, Specific Gravity Bottles, Class A
Rankem®, Specific Gravity Bottles, Class A
공급사:   RANKEM
Rankem®, Specific Gravity Bottles, Class A
공급사:  RANKEM
Certificates
Rankem®, Specific Gravity Bottles, Class A
2 Options Available Below
Product Details & Documents
Specific Gravity Bottle Class A with Ground in Thermometer range 0 to 50 °C with work Certificate (Pyknometer). Suitable for determining the density with built in thermometer.
Product Family Options
제품 정보
  • 재고 여부
    Price
  • RANKBSG-PST-0025-A
    Specific gravity bottle, 25 ml
    Specific gravity bottle, 25 ml
    Log in 재고 확인
    Log in 재고 확인
    1 * 5 items
    1 * 5 items
  • RANKBSG-PST-0050-A
    Specific gravity bottle, 50 ml
    Specific gravity bottle, 50 ml
    Log in 재고 확인
    Log in 재고 확인
    1 * 5 items
    1 * 5 items