Rankem®, Solution Bottles, Plain, Tooled Neck
공급사: RANKEM
Jump to:
이 제품에 관하여
5 Options Available Below
Product Details & Documents
Product Family Options
제품 정보
- 용량Colour재고 여부Price
- RANKBS-PTN-00250Solution bottle, O.D×H: 70×140 mm
- RANKBS-PTN-00500Solution bottle, O.D×H: 86×180 mm
- RANKBS-PTN-01000Solution bottle, O.D×H: 101×230 mm
- RANKBS-PTN-10000Solution bottle, O.D×H: 235×430 mm
- RANKBS-PTN-20000Solution bottle, O.D×H: 299×530 mm