Avantor® Alltima HP Silica, HPLC Columns
공급사: Avantor
인증서
이 제품에 관하여
Avantor® Alltima HP columns features for general purpose normal-phase applications.
- High resolution for complex samples
- High purity silica
- Excellent column stability
These columns combine the selectivity and performance needed to overcome the most challenging separation needs. The low column bleed makes these columns ideal for microbore applications.
사양
- Catalog No:
- HICH87802
- 87803/N
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Endcapped:
- No
- No
- Surface area:
- 450 m²/g
- 450 m²/g
- USP code:
- L3
- L3
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)