LiChrospher® RP-8, HPLC Columns
공급사: Avantor
인증서
이 제품에 관하여
LiChrospher® is a silica available in both endcapped and non-endcapped C8 and C18 phases.
- Spherical porous silica
- High efficiency packed columns
사양
- Catalog No:
- LISPRP8-5125AF
- LISPRP8510C5SK
- LISPRP8-5-10C5
- LISPRP8-5-125A
- LISPRP8-5-250A
- LISPRP8-5250AF
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 12.5%
- 12,5%
- 12.5%
- 12.5%
- 12.5%
- 12.5%
- Endcapped:
- No
- No
- No
- No
- No
- No
- Surface area:
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- 350 m²/g
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)