Zorbax C8, HPLC Columns
이 제품에 관하여
Zorbax spherical particles are manufactured from small extremely uniform colloidal silica micro beads which are agglutinated in a Du Pont patented organic polymerisation process. The organic polymer is removed by a controlled sintering process to yield mechanically stable silica particles.
- Spherical porous silica
- Narrow particles and pore size distribution
- Robust with good batch to batch reproducibility
: Other dimensions columns and guard cartridges are available on request.
7 Options Available Below
Product Details & Documents
Zorbax spherical particles are manufactured from small extremely uniform colloidal silica micro beads which are agglutinated in a Du Pont patented organic polymerisation process. The organic polymer is removed by a controlled sintering process to yield mechanically stable silica particles.
- Spherical porous silica
- Narrow particles and pore size distribution
- Robust with good batch to batch reproducibility
: Other dimensions columns and guard cartridges are available on request.
Specifications for Product Family
사양
- Catalog No:
- ZC8-150AS
- ZC8-150AF
- ZC8-250AF
- ZC8-150A
- ZC8-200A
- ZC8-250A
- ZC8-10C5
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- 12%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Surface area:
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- 330 m²/g
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
사양
Product Family Options
Zorbax Medium bore columns
제품 정보
- Phase컬럼 형식기공 크기입자 크기내부 직경길이재고 여부Price
- 사양:
Low Manufacturing Impact More
Specifications
Cat. No.ZC8-150ASEnvironmentally PreferableLow Manufacturing ImpactCarbon load12%EndcappedYesSurface area330 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact
Zorbax Analytical columns
제품 정보
- Phase컬럼 형식기공 크기입자 크기내부 직경길이재고 여부Price
- 사양:
Low Manufacturing Impact More
Specifications
Cat. No.ZC8-150AFEnvironmentally PreferableLow Manufacturing ImpactCarbon load12%EndcappedYesSurface area330 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - 사양:
Low Manufacturing Impact More
Specifications
Cat. No.ZC8-250AFEnvironmentally PreferableLow Manufacturing ImpactCarbon load12%EndcappedYesSurface area330 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - 사양:
Low Manufacturing Impact More
Specifications
Cat. No.ZC8-150AEnvironmentally PreferableLow Manufacturing ImpactCarbon load12%EndcappedYesSurface area330 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - 사양:
Low Manufacturing Impact More
Specifications
Cat. No.ZC8-200AEnvironmentally PreferableLow Manufacturing ImpactCarbon load12%EndcappedYesSurface area330 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - 사양:
Low Manufacturing Impact More
Specifications
Cat. No.ZC8-250AEnvironmentally PreferableLow Manufacturing ImpactCarbon load12%EndcappedYesSurface area330 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact
Zorbax Guard cartridges
제품 정보
- Phase컬럼 형식기공 크기입자 크기내부 직경길이재고 여부Price
- 사양:
Low Manufacturing Impact More
Specifications
Cat. No.ZC8-10C5Environmentally PreferableLow Manufacturing ImpactCarbon load12%EndcappedYesSurface area330 m²/gEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact