About this item
Avantor® Hichrom RPB is a high purity base deactivated silica which offers a unique selectivity due to the proprietary C8/C18 multi-alkyl bonding and exhaustive endcapping employed. The range includes 3,5, 5 and 10 μm phases, which are suitable for LC-MS applications and preparative separations respectively.
- Increased productivity due to reduced analysis time
- Decreased column length without sacrificing efficiency
- Excellent chromatography
Avantor® Hichrom RPB is specifically designed for the reversed-phase separation of basic molecules. Residual acidic sites have been eliminated whilst monolayer coverage of the high purity silica surface has been retained to obtain optimum plate efficiency. Both octyl- and octadecylsilanes are bonded to give a material which combines the robustness of a C18 phase with the high coverage of a C8 phase. Excellent peak shapes are obtained for basic drug molecules. In addition, acids and bases can be similarly chromatographed under suitably buffered conditions.
* Use with freestanding holder HI-161 and column coupler HI-081.
Specifications
- Catalog No:
- HIRPB-250P
- HI-10RPB-250P3
- HIRPB-100P3
- HIRPB-100P
- HIRPB-250SP
- HI-3.5RPB150AS
- HIRPB-10C5
- HI-10RPB-100P
- HI-3.5RPB100AF
- HIRPB-250AM
- HIRPB-100A
- HI-10RPB-50P3
- HIRPB-250AS
- HIRPB-50P3
- HIRPB-100AS
- HIRPB-250P3
- HI-3.5RPB100AS
- HI-10RPB-50SP1
- HIRPB-150P3
- HI3.5RPB10CM5
- HIRPB-150AS
- HIRPB-150P
- HIRPB-250A
- HI-3.5RPB150AM
- HIRPB-10C5-SK
- HICHHIRPB-10CP
- HIRPB-50P
- HICHHI-10RPB-10CP
- HI-10RPB-100SP1
- HI-10RPB-100P3
- HI-10RPB-150SP1
- HIRPB-150A
- HI-10RPB-250SP1
- HI-10RPB-150P
- HI-10RPB-150P3
- HI-10RPB-10CP3
- HI-10RPB-250P
- HI-10RPB-50P
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- 14%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Surface area:
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)