Orders LinkContactUsLinkComponent
Safranine O solution
Safranine O solution
공급사:  VWR Chemicals
Certificates
Safranine O solution
Safranine O solution
2 Options Available Below
Product Details & Documents
동의어:  CI 50240
  • CAS Number:477-73-6
  • 화학식:C₂₀H₁₉ClN₄
  • MDL 번호:MFCD00011759
  • Molecular Weight:350.85 g/mol
  • EINECS:207-518-8
  • UN:0000