VWR®, Waugh forceps
203 mm
제품 번호 VWRU82027-428
공급사: VWR Collection
Jump to:
Product Details & Documents
Waugh-type forceps with 1×2 teeth and guide pin.
- Stainless steel construction
- Teeth help ensure a firm grip
Forceps are straight and feature cross-serrated tips.
Specifications
- 길이:203 mm