VWR® SuperSlips™, Micro Cover Glasses, Rectangular
Supplier: VWR Collection
Jump to:
About this item
These cover glasses are corrosion-resistant and uniformly thin and flat.
- Suitable for humid environments
- Resistant to corrosion
- Processing techniques result in superior cleanliness
SuperSlip coverslips combine specialised production methods with moisture-resistant packaging to prevent sticking.
2 Options Available Below
Product Details & Documents
These cover glasses are corrosion-resistant and uniformly thin and flat.
- Suitable for humid environments
- Resistant to corrosion
- Processing techniques result in superior cleanliness
SuperSlip coverslips combine specialised production methods with moisture-resistant packaging to prevent sticking.