VWR® VisiScope® ML113 POL / BL113 POL Polarising Microscopes
공급사: VWR Collection
인증서
이 제품에 관하여
These polarising microscopes with rechargeable battery for very long outdoor operation (up to 15 hours) are ideal for educational purposes.
- Energy efficient LED illumination with light intensity control for constant pure-white colour temperature
- Coaxial coarse and fine focusing mechanism with limit stop to prevent contact between objective and specimen
- Adjustable tension of coarse focusing knob
- Rotatable round stage, 120 mm diameter
- N-plan objectives with anti-fungus treatment
- Cordless operation thanks to rechargeable Li-Ion battery (charging time: 6 hours)
납품 정보: Supplied ready to use with eyepieces, objectives, dust cover and external power supply with EU, UK and US plug for charging the Li-Ion battery. Also includes a rotating polariser (swing-out) and a fixed analyser (sliding-out).
사양
- Catalog No:
- 630-2977
- 630-2976
- Observation method:
- Brightfield and polarisation
- Brightfield and polarisation
- Head:
- 30° inclined, 360° rotating
- 30° inclined, 360° rotating
- Eyepieces:
- WF 10×/18 mm
- WF 10×/18 mm
- Nosepiece:
- Quadruple
- Quadruple
- Objectives:
- N-plan achromatic 4×, 10×, 40×
- N-plan achromatic 4×, 10×, 40×
- Stage:
- Round, Ø 120 mm
- Round, Ø 120 mm
- Condenser:
- Fixed, 1.25, pre-centered, with iris diaphragm
- Fixed, 1.25, pre-centered, with iris diaphragm
- Focusing system:
- Coaxial coarse and fine knobs
- Coaxial coarse and fine knobs
- Illumination:
- LED (white, 1 W)
- LED (white, 1 W)
사양
Eyepieces